Skip to Content
Merck
All Photos(1)

Key Documents

757527

Sigma-Aldrich

Sodium hydroxide

ultra dry, powder or crystals, 99.99% trace metals basis

Synonym(s):

‘Caustic soda’

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.99% trace metals basis
form:
powder or crystals
solubility:
water: soluble 1260 g/L at 20 °C
Pricing and availability is not currently available.

vapor density

>1 (vs air)

Quality Level

vapor pressure

<18 mmHg ( 20 °C)
3 mmHg ( 37 °C)

Assay

99.99% trace metals basis

form

powder or crystals

impurities

≤150.0 ppm Trace Metal Analysis

mp

318 °C (lit.)

solubility

water: soluble 1260 g/L at 20 °C

density

2.13 g/cm3

SMILES string

[OH-].[Na+]

InChI

1S/Na.H2O/h;1H2/q+1;/p-1

InChI key

HEMHJVSKTPXQMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Konstantinos A Polyzos et al.
Cardiovascular research, 106(2), 295-302 (2015-03-10)
Atherosclerosis is a chronic inflammatory disease that is initiated by the retention and accumulation of low-density lipoprotein in the artery, leading to maladaptive response of cells from the immune system and vessel wall. Strong evidence implicates indoleamine 2,3-dioxygenase (IDO), the
Virginie Mariot et al.
Annals of neurology, 78(3), 387-400 (2015-05-29)
Facioscapulohumeral muscular dystrophy (FSHD) is linked to either contraction of D4Z4 repeats on chromosome 4 or to mutations in the SMCHD1 gene, both of which result in the aberrant expression of the transcription factor DUX4. However, it is still difficult
C Berger et al.
Human reproduction (Oxford, England), 30(4), 800-811 (2015-03-06)
Does ulipristal acetate (UPA) used for emergency contraception (EC) interfere with the human embryo implantation process? UPA, at the dosage used for EC, does not affect human embryo implantation process, in vitro. A single pre-ovulatory dose of UPA (30 mg)
Jesper Hjortnaes et al.
Advanced healthcare materials, 4(1), 121-130 (2014-06-25)
Three dimensional (3D) hydrogel platforms are powerful tools, providing controllable, physiologically relevant microenvironments that could aid in understanding how various environmental factors direct valvular interstitial cell (VIC) phenotype. Continuous activation of VICs and their transformation from quiescent fibroblast to activated
Richard J Travers et al.
Blood, 124(22), 3183-3190 (2014-09-10)
Polyphosphate (polyP) is secreted by activated platelets and has been shown to contribute to thrombosis, suggesting that it could be a novel antithrombotic target. Previously reported polyP inhibitors based on polycationic substances, such as polyethylenimine, polyamidoamine dendrimers, and polymyxin B

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service