Skip to Content
Merck
All Photos(4)

Key Documents

C2875

Sigma-Aldrich

Octanoic acid

≥99%

Synonym(s):

Acid C8, Caprylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6COOH
CAS Number:
Molecular Weight:
144.21
Beilstein:
1747180
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

biological source

Elaeis guineensis

Quality Level

vapor density

5 (vs air)

vapor pressure

1 mmHg ( 78 °C)

Assay

≥99%

form

liquid

refractive index

n20/D 1.428 (lit.)

bp

237 °C (lit.)

mp

15-17 °C (lit.)

density

0.91 g/mL at 25 °C (lit.)

functional group

carboxylic acid

lipid type

saturated FAs

storage temp.

room temp

SMILES string

CCCCCCCC(O)=O

InChI

1S/C8H16O2/c1-2-3-4-5-6-7-8(9)10/h2-7H2,1H3,(H,9,10)

InChI key

WWZKQHOCKIZLMA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Octanoic acid is an eight carbon fatty acid, which is found in palm, coconut oil and milk. It is used to treat candidiasis and bacterial infections. Octanoic acid has antibacterial and antifungal properties. It is used in nutritional supplementation.[1]

Application

Octanoic acid has been used:
  • as a precipitating agent for the purification of antibodies[2][3](2)(4)
  • to accelerate β-oxidation for the formation of acetyl-coA akin[4]
  • to increase the rate of astrocyte ketogenesis[5]
  • in chemical 1,1′-carbonyldiimidazole (CDI) coupling of carboxylic acids to coenzyme A (CoA)[6]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1C

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Medium-chain fatty acids inhibit mitochondrial metabolism in astrocytes promoting astrocyte-neuron lactate and ketone body shuttle systems
Thevenet J, et al.
Faseb Journal, 30(5), 1913-1926 (2016)
Combining Promiscuous Acyl-CoA Oxidase and Enoyl-CoA Carboxylase/Reductases for Atypical Polyketide Extender Unit Biosynthesis
Vogeli B, et al.
Cell Chemical Biology (2018)
Natural Standard Herb & Supplement Guide - E-Book: An Evidence-Based (2016)
Oleg V Pyankov et al.
Scientific reports, 7, 41537-41537 (2017-02-06)
Herein we describe production of purified equine IgG obtained from horses immunized with plasmid DNA followed by boosting with Kunjin replicon virus-like particles both encoding a modified Ebola glycoprotein. Administration of the equine IgG over 5 days to cynomolgus macaques
Kang Cui et al.
Angewandte Chemie (International ed. in English), 59(33), 14049-14053 (2020-05-12)
Reported here is a molecular dipole that self-assembles into highly ordered patterns at the liquid-solid interface, and it can be switched at room temperature between a bright and a dark state at the single-molecule level. Using a scanning tunneling microscope

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service