Skip to Content
Merck
All Photos(1)

Key Documents

03-4560

Sigma-Aldrich

1-Butanol

JIS special grade, ≥99.0%

Synonym(s):

n-Butanol, Butyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3OH
CAS Number:
Molecular Weight:
74.12
Beilstein:
969148
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

grade

JIS special grade

vapor density

2.55 (vs air)

Assay

≥99.0%

form

liquid

autoignition temp.

649 °F

expl. lim.

11.2 %

availability

available only in Japan

refractive index

n20/D 1.399 (lit.)

bp

116-118 °C (lit.)

mp

−90 °C (lit.)

density

0.81 g/mL at 25 °C (lit.)

SMILES string

CCCCO

InChI

1S/C4H10O/c1-2-3-4-5/h5H,2-4H2,1H3

InChI key

LRHPLDYGYMQRHN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

84.2 °F

Flash Point(C)

29 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prasenjit Seal et al.
The journal of physical chemistry. A, 117(2), 275-282 (2012-12-19)
In the present work, we study the H atom abstraction reactions by hydroxyl radical at all five sites of 1-butanol. Multistructural variational transition state theory (MS-VTST) was employed to estimate the five thermal rate constants. MS-VTST utilizes a multifaceted dividing
Jie Yao et al.
Nature communications, 5, 5670-5670 (2014-11-29)
Layer-structured two-dimensional nanomaterials are a family of materials with strong covalent bonding within layers and weak van der Waals interaction between layers, whose vertical thickness can be thinned down to few nanometer and even single atomic layer. Bismuth chalcogenides are
Joel G Davis et al.
Nature, 491(7425), 582-585 (2012-11-23)
Hydrophobic hydration is considered to have a key role in biological processes ranging from membrane formation to protein folding and ligand binding. Historically, hydrophobic hydration shells were thought to resemble solid clathrate hydrates, with solutes surrounded by polyhedral cages composed
A L Gainer et al.
Clinica chimica acta; international journal of clinical chemistry, 123(1-2), 11-17 (1982-08-04)
Neutrophils were isolated in good yield from fresh whole blood and their alkaline phosphatase was solubilized. Inhibitor studies using L-phenylalanylglycylglycine, L-phenylalanine and L-homoarginine revealed a distinct pattern of inhibition for each of the crude or purified preparations of the human
R W Wulkan et al.
The International journal of biochemistry, 18(11), 1045-1051 (1986-01-01)
The butanol extraction method of Morton (1950), a routine step in enzyme purification, is discussed with special reference to a hydrophobic form of alkaline phosphatase from human liver tissue. This form slowly precipitates from butanol-extracted liver tissue homogenates stored at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service