Merck
  • Determination of organochlorines, polychlorinated biphenyls and polybrominated diphenyl ethers in human hair: estimation of external and internal exposure.
Determination of organochlorines, polychlorinated biphenyls and polybrominated diphenyl ethers in human hair: estimation of external and internal exposure.
Chemosphere (2014-08-13)
Dasheng Lu, Chao Feng, Yuanjie Lin, Dongli Wang, Ho Sai Simon Ip, Xinlei Qiu, Guoquan Wang, Jianwen She
ABSTRACT
A novel method was developed for the analysis of organochlorine pesticides (OCPs), polychlorinated biphenyls (PCBs) and polybrominated diphenyl ethers (PBDEs) in human hair samples. External contaminants of hair were extracted with acetone under sonication, while washed hair was further hydrolyzed in formic acid and acetone (1:4, v/v) with microwave assisted extraction (MAE) for internal contaminant measurements. Both internal and external extracts were cleaned up with gel permeation chromatography (GPC) and then solid phase extraction (SPE), before analyzed by a large volume injection-gas chromatography-tandem mass spectrometry (LVI-GC-MS/MS) using triple quadruple mass analyzer. Good linearity (R(2)⩾ 0.996) was established within a concentration range between 0.1 and 100 ng mL(-)(1) among all target analytes. The method was validated for accuracy, precision and sensitivity. The developed method is intended to be cost effective and robust for the routine human hair analysis of PCBs, PBDEs and OCPs including acid-labile OCPs. The described method has been applied in pilot biomonitoring study and the preliminary data suggested that the contaminant profiles with the use of partial least-squares analysis discriminant analysis (PLA-DA) could be useful in differentiating external and internal exposure.
Sigma-Aldrich
Ammonium formate, SAJ first grade, ≥95.0%
Supelco
Calcium formate, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Thallium(I) formate, 97%
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Cesium formate, 98%
Sigma-Aldrich
Calcium formate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Supelco
Ammonium formate solution, 10 mM in H2O, suitable for HPLC
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Sodium formate, 99.998% trace metals basis
Sigma-Aldrich
Potassium formate, ReagentPlus®, 99%
Sigma-Aldrich
Sodium formate-13C, 99 atom % 13C
Sigma-Aldrich
Potassium formate, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Formic acid, JIS special grade, ≥98.0%
Sigma-Aldrich
Sodium formate, BioUltra, ≥99.0% (NT)
Supelco
Sodium formate, analytical standard
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Sodium formate, reagent grade, 97%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Sodium formate, ACS reagent, ≥99.0%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%