Skip to Content
Merck
All Photos(5)

Key Documents

156264

Sigma-Aldrich

Ammonium formate

reagent grade, 97%

Synonym(s):

Formic acid ammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCO2NH4
CAS Number:
Molecular Weight:
63.06
Beilstein:
3625095
EC Number:
MDL number:
UNSPSC Code:
12352106
eCl@ss:
39021903
PubChem Substance ID:
NACRES:
NA.21
Assay:
97%
grade:
reagent grade
form:
granular
moist crystals
Pricing and availability is not currently available.

grade

reagent grade

Quality Level

Assay

97%

form

granular
moist crystals

pH

5.5-7.5 (25 °C, 63.1 g/L)

mp

119-121 °C (lit.)

density

1.26 g/mL at 25 °C (lit.)

SMILES string

N.OC=O

InChI

1S/CH2O2.H3N/c2-1-3;/h1H,(H,2,3);1H3

InChI key

VZTDIZULWFCMLS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ammonium formate is an ammonium salt of formic acid. It is a cheap and promising N-formylating agent for various amines (secondary and aniline).[1] Its anhydrous form may be employed as a catalyst for the synthesis of various amino derivatives from aliphatic and aromatic compounds.[2]
Ammonium formate can be used as a source to produce ammonia.[3] It can also be used in the reductive alkylation of amines.[4]

Application

Ammonium formate may be employed in the following studies:
  • Deprotection of N-benzyl derivatives of amino acids and amines to afford free amino acids and amines.[5]
  • Conversion of α, β-unsaturated nitroalkenes to the corresponding oximes.[6]
  • Preparation of nitriles.[7]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Copper-Catalyzed Synthesis of Nitriles by Aerobic Oxidative Reaction of Alcohols and Ammonium Formate.
Yadav DKT and Bhanage BM.
European Journal of Organic Chemistry, 23, 5106-5110 (2013)
Trost.MB
Comprehensive Organic Synthesis, 8 (1991)
Appukkuttan P et al.
Microwave Methods in Organic Synthesis, 266 (2006)
The palladium assisted transfer reduction of a, ?-unsaturated nitroalkenes to oximes using ammonium formate.
Kabalka GW, et al.
Synthetic Communications, 20(16), 2453-2458 (1990)
Rapid debenzylation of N-benzylamino derivatives to amino-derivatives using ammonium formate as catalytic hydrogen transfer agent.
Ram S and Spicer LD.
Tetrahedron Letters, 28(5), 515-516 (1987)

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. Hello, I need the expiry date of the ammonium formiate CAS 156264, batch MKCQ2722. In the COA available and in the bottle it does not have.

    1 answer
    1. Please see the Certificate of Analysis for Product 156265 Lot MKCQ2722:
      https://www.sigmaaldrich.com/certificates/COFA/15/156264/156264-BULK_______MKCQ2722__.pdf.

      This product is not assigned an expiration date or recommended retest date. Products with no expiration date or recommended retest date should be routinely inspected by customers to ensure they perform as expected. These products are also subject to a one year warranty from the date of shipment. For more information you may access the "Product Dating Information" document under "ADDITIONAL USEFUL DOCUMENTS ABOUT OUR PRODUCTS" at the bottom of the Quality Services page with this link: https://www.sigmaaldrich.com/life-science/quality-and-regulatory-management/quality-services.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service