Skip to Content
Merck
All Photos(1)

Key Documents

11-0780

Sigma-Aldrich

Formic acid

JIS special grade, ≥98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCOOH
CAS Number:
Molecular Weight:
46.03
Beilstein:
1209246
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

JIS special grade

vapor density

1.6 (vs air)

vapor pressure

44.8 mmHg ( 20 °C)

Assay

≥98.0%

form

liquid

autoignition temp.

1004 °F

expl. lim.

57 %

availability

available only in Japan

refractive index

n20/D 1.370 (lit.)

pH

2.2 (20 °C, 2.2 g/L)

bp

100-101 °C (lit.)

mp

8.2-8.4 °C (lit.)

density

1.22 g/mL at 25 °C (lit.)

SMILES string

OC=O

InChI

1S/CH2O2/c2-1-3/h1H,(H,2,3)

InChI key

BDAGIHXWWSANSR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

121.1 °F - closed cup

Flash Point(C)

49.5 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Liesivuori et al.
Pharmacology & toxicology, 69(3), 157-163 (1991-09-01)
Metabolism of methanol, methyl ethers, esters and amides give rise to formic acid. This acid is an inhibitor of the mitochondrial cytochrome oxidase causing histotoxic hypoxia. Formic acid is a weaker inhibitor than cyanide and hydrosulphide anions. The body burden
Tasqiah Julianti et al.
Journal of ethnopharmacology, 155(1), 426-434 (2014-06-04)
Leaf decoctions of Carica papaya have been traditionally used in some parts of Indonesia to treat and prevent malaria. Leaf extracts and fraction have been previously shown to possess antiplasmodial activity in vitro and in vivo. Antiplasmodial activity of extracts
Yoann Rombouts et al.
Annals of the rheumatic diseases, 74(1), 234-241 (2013-10-10)
To determine whether anticitrullinated protein antibodies (ACPA) exhibit specific changes in Fc glycosylation prior to the onset of arthritis. Serum samples of patients with ACPA-positive arthralgia (n=183) were collected at baseline and at various time points of follow-up. 105 patients
Emma Granqvist et al.
The New phytologist, 207(3), 551-558 (2015-05-27)
Plants that form root-nodule symbioses are within a monophyletic 'nitrogen-fixing' clade and associated signalling processes are shared with the arbuscular mycorrhizal symbiosis. Central to symbiotic signalling are nuclear-associated oscillations in calcium ions (Ca(2+) ), occurring in the root hairs of
Victoria G Dunne et al.
NMR in biomedicine, 18(1), 24-33 (2004-09-30)
Following subarachnoid haemorrhage the most significant complication is sustained cerebral vascular contraction (vasospasm), which may result in terminal brain damage from cerebral infarction. Despite this, the biochemical cause of vasospasm remains poorly understood. In this study, the global high-concentration metabolite

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service