Skip to Content
Merck
All Photos(2)

Key Documents

310042

Sigma-Aldrich

Thallium(I) formate

97%

Synonym(s):

Formic acid thallium(I) salt, Thallous formate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCO2Tl
CAS Number:
Molecular Weight:
251.42
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

reaction suitability

core: thallium
reagent type: catalyst

SMILES string

[Tl]OC=O

InChI

1S/CH2O2.Tl/c2-1-3;/h1H,(H,2,3);/q;+1/p-1

InChI key

UESISTHQAYQMRA-UHFFFAOYSA-M

Application

Catalyst for cyclization reactions

Used in Cherenkov radiation material

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sheng Feng et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 915-916, 28-32 (2013-01-15)
This paper described a sensitive and rapid method based on ultra-performance liquid chromatography coupled to tandem mass spectrometry (UPLC-MS/MS) for the determination of phenylephrine in human plasma. Plasma samples were pre-purified by solid-phase extraction (SPE). The chromatographic separation was achieved
Manoj Nimbalkar et al.
Journal of magnetic resonance (San Diego, Calif. : 1997), 228, 16-31 (2013-01-22)
We present highly robust, optimal control-based shaped pulses designed to replace all 90° and 180° hard pulses in a given pulse sequence for improved performance. Special attention was devoted to ensuring that the pulses can be simply substituted in a
Ji Ye Lee et al.
Bioresource technology, 132, 84-90 (2013-02-12)
The objective of this work was to investigate the feasibility of acid-catalyzed hydrothermal fractionation for maximum solubilization of the hemicellulosic portion of three agricultural residues. The fractionation conditions converted into combined severity factor (CS) in the range of 1.2-2.9. The
Lydia Beyer et al.
Journal of bacteriology, 195(7), 1428-1435 (2013-01-22)
Enterobacteria such as Escherichia coli generate formate, lactate, acetate, and succinate as major acidic fermentation products. Accumulation of these products in the cytoplasm would lead to uncoupling of the membrane potential, and therefore they must be either metabolized rapidly or
Pavel Kubáň et al.
Analytica chimica acta, 768, 82-89 (2013-03-12)
A simple method using direct injection of human blood samples and quantitative analysis of formate was developed for rapid diagnosis of methanol poisoning. A sample pretreatment device including a 500Da molecular weight cut-off dialysis membrane was in-line coupled to capillary

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service