Skip to Content
Merck
All Photos(2)

Key Documents

F0507

Sigma-Aldrich

Formic acid

reagent grade, ≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCOOH
CAS Number:
Molecular Weight:
46.03
Beilstein:
1209246
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21

grade

reagent grade

Quality Level

vapor density

1.6 (vs air)

vapor pressure

44.8 mmHg ( 20 °C)

Assay

≥95%

form

liquid

autoignition temp.

1004 °F

contains

<2.5% water as stabilizer

expl. lim.

57 %

impurities

≤1% acetic acid

refractive index

n20/D 1.370 (lit.)

pH

2.2 (20 °C, 2.2 g/L)

bp

100-101 °C (lit.)

mp

8.2-8.4 °C (lit.)

density

1.22 g/mL at 25 °C (lit.)

storage temp.

room temp

SMILES string

OC=O

InChI

1S/CH2O2/c2-1-3/h1H,(H,2,3)

InChI key

BDAGIHXWWSANSR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Formic acid (HCOOH, FA) is the simplest carboxylic acid that is mostly found in insect bites and stings. It is widely utilized as a hydrogen storage molecule due to its low toxicity, recyclability, ease of usage, and liquid state in ambient conditions.

Application


  • Electrochemical CO(2) Reduction on Metallic and Oxidized Tin: This study uses grand-canonical density functional theory (DFT) and in situ attenuated total reflectance surface-enhanced infrared absorption spectroscopy (ATR-SEIRA) to investigate electrochemical carbon dioxide reduction on tin surfaces, where formic acid could play a role in understanding reaction mechanisms (Whittaker et al., 2024).

  • Simultaneous Measurement of COVID-19 Treatment Drugs: This research demonstrates the use of UPLC-MS/MS for the simultaneous measurement of COVID-19 treatment drugs in rat plasma, indicating the importance of formic acid in preparing samples or as a mobile phase additive for better chromatographic separation (Zhou et al., 2024).

  • Metabolite Profiling of Liquiritin: The study involves metabolite profiling using ultra-high-performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS), where formic acid is likely utilized in sample preparation or chromatographic processes (Chen et al., 2024).

  • Analysis of Cocaine and Its Metabolites: This article explores solid-phase extraction followed by UHPLC-ESI-MS/MS analysis of cocaine metabolites, a method that often incorporates formic acid to enhance the ionization of analytes (Makhdoom et al., 2024).

  • Determination of Antimicrobial Compounds in Pigs: This research uses UHPLC-MS/MS for the simultaneous determination of various antimicrobial compounds, demonstrating formic acid′s role in sample processing and chromatographic separation (Nowacka-Kozak et al., 2024).

Formic acid is used as a reducing agent in the:

  • synthesis of graphene from graphene oxide.
  • catalytic reduction of chromium (Cr(VI) to Cr(III)) by colloidal palladium.
It is also used as a reagent in the Eschweiler-Clarke reaction during the synthesis of ketamine from the reduction of 2-(2-chlorophenyl)-2-nitrocyclohexanone (2-CPNCH) to norketamine.

Additionally, it is used as a hydrogen donor during the transformation of the furanose form into the pyranose form of glucose and in the catalytic transfer hydrogenation reaction.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

121.1 °F - closed cup

Flash Point(C)

49.5 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new process of ketamine synthesis from 2-(2-chlorophenyl)-2-nitrocyclohexanone proposed by analyzing drug materials and chemicals seized in Taiwan
Yao-Te Y, et al.
Forensic Science International, 349, 111776-111776 (2023)
Palladium nanoparticles for catalytic reduction of Cr (VI) using formic acid
Omole, Marcells A O, et al.
Applied Catalysis. B, Environmental, 76, 158-167 (2007)
Catalytic transfer hydrogenation of sugar derivatives
Laszlo J, et al.
Carbohydrate Polymers, 45, 139-145 (2001)
Tissue distribution, effects of cooking and parameters affecting the extraction of azaspiracids from mussels, Mytilus edulis, prior to analysis by liquid chromatography coupled to mass spectrometry.
Hess P et al.
Toxicon, 46, 62-62 (2005)
Glucosinolates determination in tissues of horseradish Plant.
Lelario F et al.
Bio-protocol, 5, e1562-e1562 (2015)

Articles

LC-MS/MS-based determination of 19 sulfonamides, 13 quinolones, and 3 tetracycline drug residues in pork meat samples using Supel™ Swift HLB for extraction.

LC-MS/MS-based determination of 19 sulfonamides, 13 quinolones, and 3 tetracycline drug residues in pork meat samples using Supel™ Swift HLB for extraction.

LC-MS/MS-based determination of 19 sulfonamides, 13 quinolones, and 3 tetracycline drug residues in pork meat samples using Supel™ Swift HLB for extraction.

LC-MS/MS-based determination of 19 sulfonamides, 13 quinolones, and 3 tetracycline drug residues in pork meat samples using Supel™ Swift HLB for extraction.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service