Skip to Content
Merck
All Photos(1)

Key Documents

48567

Supelco

Anthracene

analytical standard

Synonym(s):

Anthraxcene, Paranaphthalene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10
CAS Number:
Molecular Weight:
178.23
Colour Index Number:
10790
Beilstein:
1905429
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

vapor density

6.15 (vs air)

vapor pressure

1 mmHg ( 145 °C)

CofA

current certificate can be downloaded

autoignition temp.

1004 °F

packaging

ampule of 5000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

340 °C (lit.)

mp

210-215 °C (lit.)

solubility

alcohols: soluble
benzene: soluble
chloroform: soluble
hydronaphthalenes: soluble
supercritical carbon dioxide: soluble

application(s)

environmental

format

neat

storage temp.

2-30°C

SMILES string

c1ccc2cc3ccccc3cc2c1

InChI

1S/C14H10/c1-2-6-12-10-14-8-4-3-7-13(14)9-11(12)5-1/h1-10H

InChI key

MWPLVEDNUUSJAV-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

General description

Anthracene is a polycyclic aromatic hydrocarbon often formed by incomplete combustion of organic materials.

Application

Anthracene has been shown to be soluble in a variety of binary and ternary mixtures of cyclohexanone, ethyl acetate, and methanol .
Anthracene may be used as an analytical reference standard for the determination of the analyte in aqueous solution by synchronous fluorimetry.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

249.8 °F - closed cup

Flash Point(C)

121.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Determination of polycyclic aromatic hydrocarbons in water samples using high-performance liquid chromatography with amperometric detection.
Nirmaier HP, et al.
Journal of Chromatography A, 730(1-2), 169-175 (1996)
Simultaneous determination of dissolved anthracene and pyrene in aqueous solution by synchronous fluorimetry.
Cai ZQ, et al.
Spectrochimica Acta. Part A, Molecular and Biomolecular Spectroscopy, 69(1), 130-133 (2008)
Payam Payamyar et al.
Chimia, 67(4), 283-285 (2013-08-24)
We describe the challenges involved with extending the limited lateral size of two-dimensional polymers (2DPs). An amphiphilic monomer with three-fold symmetry is chosen to form an ideally tessellated monolayer at the air/water interface. Anthracene [4+4] photo-dimerization is chosen as the
Hyunjung Lee et al.
Inorganic chemistry, 51(20), 10904-10915 (2012-09-26)
The tendency of a Hg(II) ion to strongly quench fluorescence of potential fluorescent sensors is explored. Fluorescence measurements show the expected order of the chelation-enhanced fluorescence (CHEF) effect of Zn(II) > Cd(II) > Hg(II) ~ Cu(II), which is interpreted as
Huan Wang et al.
Physical chemistry chemical physics : PCCP, 15(10), 3527-3534 (2013-02-05)
The orientation factor κ(2) ranging from 0 to 4, which depends on the relative orientation of the transition dipoles of the energy donor (D) and the energy acceptor (A) in space, is one of the pivotal factors deciding the efficiency

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service