Skip to Content
Merck
All Photos(3)

Key Documents

A89200

Sigma-Aldrich

Anthracene

reagent grade, 97%

Synonym(s):

Anthraxcene, Paranaphthalene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10
CAS Number:
Molecular Weight:
178.23
Colour Index Number:
10790
Beilstein:
1905429
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39011608
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

grade

reagent grade

Quality Level

vapor density

6.15 (vs air)

vapor pressure

1 mmHg ( 145 °C)

Assay

97%

form

flakes
powder or solid

autoignition temp.

1004 °F

bp

340 °C (lit.)

mp

210-215 °C (lit.)

solubility

alcohols: soluble
benzene: soluble
chloroform: soluble
hydronaphthalenes: soluble
supercritical carbon dioxide: soluble

SMILES string

c1ccc2cc3ccccc3cc2c1

InChI

1S/C14H10/c1-2-6-12-10-14-8-4-3-7-13(14)9-11(12)5-1/h1-10H

InChI key

MWPLVEDNUUSJAV-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

General description

Anthracene, a polycyclic aromatic hydrocarbon, is an important moiety in supramolecular chemistry due to its size and shape and photophysical properties. Its closed-shelled Huckel aromatic structure, fluorescence property, well-resolved absorption and emission bands, high fluorescence quantum yield and nanosecond lifetime makes it an ideal organic fluorophore.[1]

Application

Anthracene has been shown to be soluble in a variety of binary and ternary mixtures of cyclohexanone, ethyl acetate, and methanol .

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

249.8 °F - closed cup

Flash Point(C)

121.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular architectures of multi-anthracene assemblies.
Yoshizawa M & Klosterman JK.
Chemical Society Reviews, 43(6), 1885-1898 (2014)
Kohei Yazaki et al.
Chemical communications (Cambridge, England), 49(16), 1630-1632 (2013-01-23)
A bowl-shaped organic host was prepared by linking two anthracene-embedded bispyridine ligands with two methylene spacers. The water-soluble host has a hemispherical hydrophobic cavity (∼1 nm in diameter) with two cationic methylenebispyridinyl (Lewis acidic) moieties and shows the selective recognition
Ling-Bao Xing et al.
Langmuir : the ACS journal of surfaces and colloids, 29(9), 2843-2848 (2013-02-12)
A new type of anthracene organogelator based on uracil was obtained using organic aromatic solvents, cyclohexane, DMSO, ethanol, and ethyl acetate. It was further characterized by field-emission scanning electron microscopy and transmission electron microscopy. Specifically, the resulting organogels were demonstrated
Jiang-Fei Xu et al.
Organic letters, 15(24), 6148-6151 (2013-11-19)
Dynamic covalent bonds supplied by reversible anthracene dimerization were combined with pillar[5]arene/imidazole host-guest interactions to construct double-dynamic polymers. Heating such polymers (in solution or as a gel) led to depolymerization by dissociation of either the host-guest complexes alone or the
Yongshu Xie et al.
Chemical communications (Cambridge, England), 48(94), 11513-11515 (2012-10-24)
2,2'-Dipyridylamine and anthracene units were linked to afford highly emissive compounds whose Cu(2+) ensembles were developed as effective fluorescence turn-on CN(-) probes.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service