Skip to Content
Merck
All Photos(1)

Key Documents

48535

Supelco

Fluoranthene

analytical standard

Synonym(s):

Benzo[j,k]fluorene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H10
CAS Number:
Molecular Weight:
202.25
Beilstein:
1907918
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

Quality Level

CofA

current certificate can be downloaded

packaging

ampule of 5000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

384 °C (lit.)

mp

105-110 °C (lit.)

application(s)

environmental

format

neat

storage temp.

2-30°C

SMILES string

c1ccc-2c(c1)-c3cccc4cccc-2c34

InChI

1S/C16H10/c1-2-8-13-12(7-1)14-9-3-5-11-6-4-10-15(13)16(11)14/h1-10H

InChI key

GVEPBJHOBDJJJI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

FA, a six ringed highly stable compound, is considered as an environmental indicator. It is mainly selected for simulation study.
Fluoranthene (FA) is a polycyclic aromatic hydrocarbon, which can be used in the preparation of fluorescent chemosensors for the specific detection of picric acid.

Application

FA was used after dissolving in HPLC grade acetonitrile, in order to determine retention data and to study the linearity of the detector, during HPLC analysis of PAH (polycyclic aromatic hydrocarbon).
Fluoranthene (FA) may also be used as an analytical standard for the determination of the analyte in food samples, oils, fats, and environmental samples by various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

388.4 °F - closed cup

Flash Point(C)

198.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Fluoranthene based fluorescent chemosensors for detection of explosive nitroaromatics
Venkatramaiah.N, et al.
Chemical Communications (Cambridge, England), 48, 5007-5009 (2012)
Estimation of polycyclic aromatic hydrocarbons associated with size segregated combustion aerosols generated from household fuels.
Tiwari, M., et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 106, 79-86 (2013)
L D Gratz et al.
Journal of hazardous materials, 74(1-2), 37-46 (2000-04-27)
For laboratories involved in polycyclic aromatic hydrocarbon (PAH) analyses in environmental samples, it is very useful to participate in interlaboratory comparison studies which provide a mechanism for comparing analytical methods. This is particularly important when PAH analyses are routinely done
Comparison of GC-MS and HPLC for overcoming matrix interferences in the analysis of PAHs in smoked food
Chiu CP, et al.
Chromatographia, 44(9-10), 497-504 (1997)
G Grimmer et al.
Journal - Association of Official Analytical Chemists, 58(4), 725-733 (1975-07-01)
A method is described for the determination of polycyclic aromatic hydrocarbons (PAHs) with 3-7 rings in (I) meat, poultry, fish, and yeast; and (II) oils and fats. The extraction of PAHs from group I is incomplete, and, therefore, group I

Articles

Highly accurate HPLC-FLD method for evaluating 15 PAHs in olive oil using Ascentis® Express PAH HPLC column and SPE cleanup, following GB 5009.265-2021.

Highly accurate HPLC-FLD method for evaluating 15 PAHs in olive oil using Ascentis® Express PAH HPLC column and SPE cleanup, following GB 5009.265-2021.

Highly accurate HPLC-FLD method for evaluating 15 PAHs in olive oil using Ascentis® Express PAH HPLC column and SPE cleanup, following GB 5009.265-2021.

Highly accurate HPLC-FLD method for evaluating 15 PAHs in olive oil using Ascentis® Express PAH HPLC column and SPE cleanup, following GB 5009.265-2021.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service