Skip to Content
Merck
All Photos(1)

Key Documents

8.20109

Sigma-Aldrich

Anthracene

for synthesis

Synonym(s):

Anthracene, Paranaphthalene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10
CAS Number:
Molecular Weight:
178.23
MDL number:
UNSPSC Code:
12352005
EC Index Number:
204-371-1
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

0.000008 hPa ( 25 °C)

Quality Level

Assay

≥98.5% (GC)

form

solid

autoignition temp.

540 °C

potency

>16000 mg/kg LD50, oral (Rat)

expl. lim.

0.6 % (v/v)

bp

340 °C/1013 hPa

mp

213-217 °C

transition temp

flash point 121 °C

solubility

<0.0001 g/L

density

1.13 g/cm3 at 20 °C

bulk density

330 kg/m3

storage temp.

2-30°C

SMILES string

c21c(cc3c(c2)cccc3)cccc1

InChI

1S/C14H10/c1-2-6-12-10-14-8-4-3-7-13(14)9-11(12)5-1/h1-10H

InChI key

MWPLVEDNUUSJAV-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.5 % (a/a)
Melting range (lower value): ≥ 213 °C
Melting range (upper value): ≤ 217 °C
Identity (IR): passes test

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

249.8 °F - Pensky-Martens closed cup

Flash Point(C)

121 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service