Skip to Content
Merck
All Photos(1)

Key Documents

P6149

Sigma-Aldrich

Progesterone

γ-irradiated, BioXtra, suitable for cell culture

Synonym(s):

4-Pregnene-3,20-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H30O2
CAS Number:
Molecular Weight:
314.46
Beilstein:
1915950
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.77
biological source:
synthetic (organic)
form:
powder

biological source

synthetic (organic)

Quality Level

description

synthesized from materials derived from plant source

sterility

γ-irradiated

product line

BioXtra

Assay

≥99%

form

powder

potency

0.1-20 ng/mL

technique(s)

cell culture | mammalian: suitable

mp

128-132 °C (lit.)

solubility

chloroform: 50 mg/mL, clear, colorless to light yellow

shipped in

ambient

storage temp.

room temp

SMILES string

CC(=O)[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C21H30O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h12,16-19H,4-11H2,1-3H3/t16-,17+,18-,19-,20-,21+/m0/s1

InChI key

RJKFOVLPORLFTN-LEKSSAKUSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Use in cell culture applications to study sterol regulation of cell signaling and gene expression.

Biochem/physiol Actions

Induces maturation and secretory activity of the uterine endothelium; suppresses ovulation.
Induces maturation and secretory activity of the uterine endothelium; suppresses ovulation. Progesterone is implicated in the etiology of breast cancer.

Other Notes

Steroid hormone produced by the corpus luteum.

Physical form

powder-RT; stock-frozen in working aliquots, avoid repeated freeze/thaw

Reconstitution

To prepare 20 μg/ml stock solution, add 1 ml absolute ethanol, gently swirl to dissolve, add 49 ml sterile medium.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Lact. - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nina Lisofsky et al.
NeuroImage, 118, 154-162 (2015-06-10)
Hippocampal volume has been shown to be sensitive to variations in estrogen and progesterone levels across rodents' estrous cycle. However, little is known about the covariation of hormone levels and brain structure in the course of the human menstrual cycle.
Rudolf Kaaks et al.
International journal of cancer, 134(8), 1947-1957 (2013-10-25)
Results from prospective studies on premenopausal serum hormone levels in relation to breast cancer risk have been inconclusive, especially with regard to tumor subtypes. Using a case-control study nested within the prospective European Prospective Investigation into Cancer and Nutrition (EPIC)
Cathrin Brisken
Nature reviews. Cancer, 13(6), 385-396 (2013-05-25)
Understanding the biology of the breast and how ovarian hormones impinge on it is key to rational new approaches in breast cancer prevention and therapy. Because of the success of selective oestrogen receptor modulators (SERMs), such as tamoxifen, and aromatase
Eszter Papp et al.
The Journal of infectious diseases, 211(1), 10-18 (2014-07-18)
Protease inhibitor (PI)-based combination antiretroviral therapy (cART) is administered during pregnancy to prevent perinatal human immunodeficiency virus (HIV) transmission. However, PI use has been associated with adverse birth outcomes, including preterm delivery and small-for-gestational-age (SGA) births. The mechanisms underlying these
Erika Comasco et al.
Human brain mapping, 35(9), 4450-4458 (2014-03-13)
Premenstrual dysphoric disorder (PMDD) is the prototypical sex-specific disorder in which symptom onset and offset require a particular hormonal milieu and for which there is moderate heritability. The present study investigated brain emotion processing in PMDD and healthy controls, as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service