Skip to Content
Merck
All Photos(1)

Key Documents

N5254

Sigma-Aldrich

Neuraminidase Agarose from Clostridium perfringens (C. welchii)

Type VI-A, ammonium sulfate suspension

Synonym(s):

Acylneuraminyl hydrolase, Receptor-destroying enzyme, Sialidase

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

type

Type VI-A

form

ammonium sulfate suspension

specific activity

20-60 units/g agarose

extent of labeling

0.6-1.8 units per mL gel
20-60 units per g agarose

matrix

beaded agarose

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Neuraminidase enzymes are glycoside hydrolase enzymes that catalyze hydrolysis of terminal sialic acid residues. The most well-known are the viral nearamidases, which promote influenza virus release.

Application

Agarose-linked neuraminidase from Clostridium perfringens can be used for the detection of total and desialylated sex hormone-binding globulin in human serum samples.
Neuraminidase from Clostridium perfringens has been used in a study to assess a genetic polymorphism of human serum glycoprotein (inter-α-trypsin-inhibitor). It has also been used in a study to investigate the inhibition of neuraminidase by chemically sulphated glycopeptides.

Unit Definition

One unit will liberate 1.0 μmole of N-acetylneuraminic acid per min at pH 5.0 at 37 °C using NAN-lactose or bovine submaxillary mucin, unless otherwise specified. Prices based on units using NAN-lactose as substrate.

Physical form

Suspension in 2.0 M (NH4)2SO4 solution, pH 7.0.

Preparation Note

Prepared from Neuraminidase, Type VI (N 3001).

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T I Arnon et al.
European journal of immunology, 31(9), 2680-2689 (2001-09-06)
Natural killer (NK) cells destroy virus-infected and tumor cells without prior antigen stimulation. The NK cell cytotoxicity is regulated in large part by the expression of NK cell receptors that are able to bind major histocompatibility complex (MHC) class I
N Mian et al.
The Biochemical journal, 181(2), 377-385 (1979-08-01)
Chemically sulphated glycopeptides (derived from pig duodenal mucosa) inhibited Clostridium perfringens neuraminidase (EC 3.2.1.18) activity in a pH-dependent manner. Analysis of inhibition kinetics data indicated that, although the enzyme inhibition could not be categorized into any of the classical types
U Vogt et al.
Human genetics, 84(2), 151-154 (1990-01-01)
A new genetic polymorphism of a human serum glycoprotein, the inter-alpha-trypsin-inhibitor (ITI), has been demonstrated by population and family studies. Sera were examined after neuraminidase treatment by isoelectric focusing on agarose gels followed by immunoblotting or by immunofixation with specific
The mechanisms controlling the recognition of tumor- and virus-infected cells by NKp46
Arnon, T.I., et al
Immunobiol., 103(2), 664-672 (2004)
Measurements of total and desialylated sex hormone binding globulin in serum by ELISA.
J Vaysse et al.
Clinical chemistry, 44(4), 882-884 (1998-04-29)

Articles

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service