Skip to Content
Merck
All Photos(1)

Key Documents

N3001

Sigma-Aldrich

Neuraminidase from Clostridium perfringens (C. welchii)

Type VI, lyophilized powder, 6-15 units/mg protein (using 4MU-NANA), 2-10 units/mg protein (mucin)

Synonym(s):

Acyl-neuraminyl Hydrolase, Receptor-destroying enzyme, Sialidase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

Clostridium perfringens str. 13

Quality Level

type

Type VI

form

lyophilized powder

specific activity

2-10 units/mg protein (mucin)
6-15 units/mg protein (using 4MU-NANA)

composition

Protein, ≥50% biuret

storage temp.

−20°C

Gene Information

Clostridium perfringens str. 13 ... nanI(988807)

Looking for similar products? Visit Product Comparison Guide

General description

Neuraminidase enzymes are hydrolase enzymes that promote influenza virus release from infected cells and facilitate virus spread.

Application

Neuraminidase from Clostridium perfringens (C. welchii) has been used in a study to assess a glycoprotein faction suitable for use as a substrate in preparation assays. It has also been used in a study to investigate the action of an epsilion-toxin on MDCK cells.

Biochem/physiol Actions

Neuraminidase cleavage of sialic acid groups has been used to study recognition by antibodies of glycoprotein structures. The use of neuraminidase in the estimation of N-acetylneuraminic acid was compared favorably to two other methods.
Neuraminidase from Clostridium perfringens reduces the viability of human leukemic myeloblasts and attenuates their ability to activate lymphocytes.
Neuraminidases are used to cleave terminal N-acetyl neuraminic acid (sialic acid) from a variety of glycoproteins. The enzyme from Clostridium perfringens cleaves terminal sialic acid residues which are α-2,3- α-2,6- or α-2,8-linked to Gal, GlcNac, GalNAc, AcNeu, GlcNeu, oligosaccharides, glycolipids or glycoproteins. The relative rate of cleavage decreases in the order: α-2-3 > α-2-6 . α-2-8. Neuraminidase from C. perfringens cleaves α-2-3 linked sialic acid residues most efficiently, compared to A. ureafaciens, (Sigma N3642) which preferentially cleaves α-2-6 linked residues.
The use of neuraminidase to remove sialic acid residues from glycoproteins on cell surfaces has been frequently reported. Generally, procedures have indicated using neuraminidase in PBS at 37°C for 30 minutes, followed by several washings with PBS. Treatment of tissue sections with neuraminidase at much lower concentrations require longer incubation: for 1-4 U/mL in 0.1 M acetate buffer pH 4.2-5, from 2 to 20 hours at 37 °C.

Preparation Note

Chromatographically purified from Type V (N 2876)

Analysis Note

Package sizes based on 4MU-NANA units
Package sizes based on the 4MU-NANA units

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A G Fraser et al.
Journal of medical microbiology, 8(2), 235-249 (1975-05-01)
A glycoprotein fraction (fraction VII) suitable for use as a substrate in assays of microbial neuraminidase was prepared from pooled human plasma. It is pasteurised during preparation to eliminate the risk of transmission of serum hepatitis. This results in polymerisation
Ana León-Rodríguez et al.
Scientific reports, 12(1), 11581-11581 (2022-07-09)
Short-term behavioral alterations are associated with infection and aid the recovery from sickness. However, concerns have raised that sustained behavioral disturbances after acute neuroinflammation could relate to neurological diseases in the long run. We aimed to explore medium- and long-term
L Petit et al.
Journal of bacteriology, 179(20), 6480-6487 (1997-10-23)
Epsilon-toxin is produced by Clostridium perfringens types B and D and is responsible for a rapidly fatal enterotoxemia in animals, which is characterized by edema in several organs due to an increase in blood vessel permeability. The Madin-Darby canine kidney
A Ulloa-Aguirre et al.
Biology of reproduction, 30(2), 382-387 (1984-03-01)
Graded removal of sialic acid residues from a purified preparation of rat follicle-stimulating hormone (FSH; NIADDK-FSH-I-5) by neuraminidase digestion resulted in the production of FSH isohormones with isoelectric points identical to those found within pituitary tissue. In addition, each neuraminidase-produced
A simple method for the purification of commercial neuraminidase preparations free from proteases.
M W Hatton et al.
Biochimica et biophysica acta, 327(1), 114-120 (1973-11-15)

Articles

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Protocols

Enzymatic Assay of Neuraminidase applies to products that have a specification for neuraminidase content by enzymatic determination.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service