Skip to Content
Merck
All Photos(4)

Key Documents

M8639

Sigma-Aldrich

2′-(4-Methylumbelliferyl)-α-D-N-acetylneuraminic acid sodium salt hydrate

fluorogenic, ≥95% (HPLC), powder

Synonym(s):

4-Methylumbelliferyl-N-acetyl-α-D-neuraminic acid sodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H24NNaO11 · xH2O
CAS Number:
Molecular Weight:
489.41 (anhydrous basis)
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

product name

2′-(4-Methylumbelliferyl)-α-D-N-acetylneuraminic acid sodium salt hydrate, ≥95% (HPLC)

Assay

≥95% (HPLC)

form

powder

solubility

water: 50 mg/mL, clear to slightly hazy, colorless to light yellow

storage temp.

−20°C

SMILES string

O.[H]C(O)(CO)[C@]([H])(O)[C@@H]1OC(C[C@H](O)[C@H]1NC(C)=O)(Oc2ccc3C(C)=CC(=O)Oc3c2)C(=O)O[Na]

InChI

1S/C21H25NO11.Na.H2O/c1-9-5-16(27)31-15-6-11(3-4-12(9)15)32-21(20(29)30)7-13(25)17(22-10(2)24)19(33-21)18(28)14(26)8-23;;/h3-6,13-14,17-19,23,25-26,28H,7-8H2,1-2H3,(H,22,24)(H,29,30);;1H2/q;+1;/p-1/t13-,14?,17+,18?,19+,21+;;/m0../s1

InChI key

NSQMRVBWXQQIKF-NVRWCLOTSA-M

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Substrate for fluorometric assay of neuraminidase. Used for fluorescent staining of sialidases in PAGE.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Y Fedorov et al.
Voprosy virusologii, 65(2), 113-118 (2020-06-10)
The classic hemagglutination inhibition reaction (RTGA) is used to determine the level of antiviral antibodies in human and animal serum specimens. During the performance of RTGA the tested sera must be treated with a receptor-destroying enzyme (RDE) to remove serum
Jian Zhang et al.
Journal of medicinal chemistry, 61(14), 6379-6397 (2018-07-03)
On the basis of our earlier discovery of N1-selective inhibitors, the 150-cavity of influenza virus neuraminidases (NAs) could be further exploited to yield more potent oseltamivir derivatives. Among the synthesized compounds, 15b and 15c were exceptionally active against both group-1
Li Zhang et al.
Viruses, 12(5) (2020-05-24)
Influenza virus infections can lead to viral pneumonia and acute respiratory distress syndrome in severe cases, causing significant morbidity and mortality and posing a great threat to human health. Because of the diversity of influenza virus strains and drug resistance
Amanda R Luu et al.
The Journal of biological chemistry, 295(39), 13556-13569 (2020-07-31)
Mutations in the galactosidase β 1 (GLB1) gene cause lysosomal β-galactosidase (β-Gal) deficiency and clinical onset of the neurodegenerative lysosomal storage disease, GM1 gangliosidosis. β-Gal and neuraminidase 1 (NEU1) form a multienzyme complex in lysosomes along with the molecular chaperone
Shuang Chen et al.
Annals of translational medicine, 8(13), 823-823 (2020-08-15)
Pharmacological induction of autophagy can protect against acetaminophen (APAP) induced acute liver failure (ALF) by removing APAP adducts (APAP-AD), but its mechanism is not well understood. Hepatoprotective effect of saponins from traditional Chinese medicine has attracted widespread attention from all

Articles

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service