Skip to Content
Merck
All Photos(3)

Key Documents

N2133

Sigma-Aldrich

Neuraminidase from Clostridium perfringens (C. welchii)

Type X, lyophilized powder, ≥50 units/mg protein (using 4MU-NANA)

Synonym(s):

Acyl-neuraminyl Hydrolase, Receptor-destroying enzyme, Sialidase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

type

Type X

Quality Level

form

lyophilized powder

specific activity

≥50 units/mg protein (using 4MU-NANA)

storage temp.

−20°C

Gene Information

Clostridium perfringens str. 13 ... nanI(988807)

Looking for similar products? Visit Product Comparison Guide

General description

Neuraminidase enzymes are glycoside hydrolase enzymes that catalyze hydrolysis of terminal sialic acid residues. The most well-known are the viral nearamidases, which promote influenza virus release.

Application

Neuraminidase from Clostridium perfringens has been used in a study to assess purification via affinity chromatography. It has also been used in a study to investigate site-directed mutations of amino acids of the neuraminidase gene, nanH.

Biochem/physiol Actions

Neuraminidase can block attachment of type 3 reovirus to cell membranes. This effect is related to the ability of neuraminidase to hydrolysis sialic acid residues within cell surface receptors.
Neuraminidase cleavage of sialic acid groups has been used to study recognition by antibodies of glycoprotein structures. The use of neuraminidase in the estimation of N-acetylneuraminic acid was compared favorably to two other methods.
Neuraminidases are used to cleave terminal N-acetyl neuraminic acid (sialic acid) from a variety of glycoproteins. The enzyme from Clostridium perfringens cleaves terminal sialic acid residues which are α-2,3- α-2,6- or α-2,8-linked to Gal, GlcNac, GalNAc, AcNeu, GlcNeu, oligosaccharides, glycolipids or glycoproteins. The relative rate of cleavage decreases in the order: α-2-3 > α-2-6 . α-2-8. Neuraminidase from C. perfringens cleaves α-2-3 linked sialic acid residues most efficiently, compared to A. ureafaciens, (Sigma N3642) which preferentially cleaves α-2-6 linked residues.
The use of neuraminidase to remove sialic acid residues from glycoproteins on cell surfaces has been frequently reported. Generally, procedures have indicated using neuraminidase in PBS at 37°C for 30 minutes, followed by several washings with PBS. Treatment of tissue sections with neuraminidase at much lower concentrations require longer incubation: for 1-4 U/mL in 0.1 M acetate buffer pH 4.2-5, from 2 to 20 hours at 37 °C.

Unit Definition

One unit will hydrolyze 1.0 micromole of 2′-(4-Methylumbelliferyl)-alpha-D-N-actetylneuraminic acid per min at pH 5.0 at 37 °C.

Preparation Note

Purified by affinity chromatography from Type VIII (N 5631)

Analysis Note

Package sizes based on 4MU-NANA units
Package sizes based on the 4MU-NANA units

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J R Gentsch et al.
Journal of virology, 56(2), 356-364 (1985-11-01)
The effect of pretreatment of murine L cells with bacterial neuraminidases on type 3 reovirus attachment was examined. We observed that such treatments resulted in a 60 to 80% decrease of subsequent attachment of 35S-labeled type 3 reovirus in a
C H Chien et al.
Enzyme and microbial technology, 19(4), 267-276 (1996-09-01)
The small nanH gene encoding the neuraminidase from Clostridium perfringens ATCC 10543 was cloned in JM109 using pUC19 as a vector. Sequence analysis revealed an ORF encoding 382 amino acids without a signal peptide sequence. Four regions of amino-acid sequence
Marjorie E Kanof
Current protocols in immunology, Chapter 7, 7-7 (2008-04-25)
This unit describes a procedure for separating T cells from other mononuclear cells by exploiting the unique ability of cells to bind to and form rosettes with sheep red blood cells (SRBC). This isolation method also allows recovery of the
Purification of neuraminidases from Vibrio Cholerae, Clostridium Perfringens and influenza virus by affinity chromatography.
P Cuatrecasas et al.
Biochemical and biophysical research communications, 44(1), 178-184 (1971-07-02)
Emi Yamaguchi et al.
Journal of gastroenterology and hepatology, 22(7), 1134-1140 (2007-07-05)
The authors' previous report revealed that endothelin-1 might be released from B lymphocytes in cirrhotic patients with hypersplenism. Other investigators have shown that persistent exposure to environmental contaminants including arsenic might induce idiopathic portal hypertension. The aim of this study

Articles

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Protocols

Enzymatic Assay of Neuraminidase applies to products that have a specification for neuraminidase content by enzymatic determination.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service