Skip to Content
Merck
All Photos(3)

Documents

M3657

Sigma-Aldrich

4-Methylumbelliferyl α-D-mannopyranoside

fluorogenic, ≥97% (HPLC), powder

Synonym(s):

4-methylumbelliferyl a-d-mannopyranoside, 4-methylumbelliferyl alpha-d-mannopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18O8
CAS Number:
Molecular Weight:
338.31
Beilstein:
1266648
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

product name

4-Methylumbelliferyl α-D-mannopyranoside, ≥97% (HPLC)

Quality Level

Assay

≥97% (HPLC)

form

powder

solubility

pyridine: 20 mg/mL, clear, colorless to faintly yellow

fluorescence

λex 350 nm; λem 375 nm (pH 7.0)
λex 360 nm; λem 449 nm (Reaction product)

storage temp.

−20°C

SMILES string

CC1=CC(=O)Oc2cc(O[C@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@@H]3O)ccc12

InChI

1S/C16H18O8/c1-7-4-12(18)23-10-5-8(2-3-9(7)10)22-16-15(21)14(20)13(19)11(6-17)24-16/h2-5,11,13-17,19-21H,6H2,1H3/t11-,13-,14+,15+,16+/m1/s1

InChI key

YUDPTGPSBJVHCN-VMMWWAARSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Methylumbelliferyl α-D-mannopyranoside has been used to assay α-mannosidase activity in various biological samples.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wang-Sik Lee et al.
The Journal of biological chemistry, 282(37), 27198-27203 (2007-07-27)
UDP-GlcNAc:lysosomal enzyme N-acetylglucosamine-1-phosphotransferase (GlcNAc-1-phosphotransferase) mediates the first step in the synthesis of the mannose 6-phosphate recognition marker on acid hydrolases. The transferase exists as an alpha(2)beta(2)gamma(2) hexameric complex with the alpha- and beta-subunits derived from a single precursor molecule. The
A J Sophianopoulos et al.
Archives of biochemistry and biophysics, 246(2), 572-580 (1986-05-01)
The stoichiometry of Mn2+ binding to concanavalin A at pH 6.4-7 which had been established in two independent studies [J.A. Sophianopoulos, A.J. Sophianopoulos, and W.C. MacMahon (1983) Arch. Biochem. Biophys. 223, 350-359; D.J. Christie, G.R. Munske, and J.A. Magnuson (1979)
H G Leusch et al.
Zentralblatt fur Bakteriologie : international journal of medical microbiology, 275(1), 118-122 (1991-04-01)
Immobilized purified CEA (carcinoembryonic antigen), NCA (non-specific crossreacting antigen) and BGP I (biliary glycoprotein I) bind strains of E. coli (including EPEC) and some Salmonella species (including S. typhi, S. paratyphi A + B and S. java) while Shigella-, Yersinia-
K S Shashidhara et al.
International journal of biological macromolecules, 44(1), 112-115 (2008-11-29)
Energetics of the catalysis of Class II alpha-mannosidase (E.C.3.2.1.24) from Aspergillus fischeri was studied. The enzyme showed Kcat/Km for Man (alpha1-3) Man, Man (alpha1-2) Man and Man (alpha1-6) Man as 7488, 5376 and 3690 M(-1) min(-1), respectively. The activation energy
J M Foster et al.
Insect biochemistry and molecular biology, 27(7), 657-661 (1997-07-01)
The alpha-mannosidases are implicated in both the catabolism of carbohydrates and the N-linked glycosylation pathway in insects, but little is known of the biochemistry of these glycosidases. In order to study the soluble alpha-mannosidases of Drosophila melanogaster we have used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service