Skip to Content
Merck
All Photos(6)

Key Documents

M3633

Sigma-Aldrich

4-Methylumbelliferyl β-D-glucopyranoside

β-glucosidase substrate, fluorogenic, ≥99% (HPLC), powder

Synonym(s):

MUD

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18O8
CAS Number:
Molecular Weight:
338.31
Beilstein:
94672
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Product Name

4-Methylumbelliferyl β-D-glucopyranoside, β-glucosidase substrate

Assay

≥99% (HPLC)

form

powder

solubility

DMF: 50 mg/mL, clear, colorless to faintly yellow

fluorescence

λex 316 nm; λem 372 nm (pH 9.1)
λex 360 nm; λem 449 nm (Reaction product)

storage temp.

−20°C

SMILES string

CC1=CC(=O)Oc2cc(O[C@@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)ccc12

InChI

1S/C16H18O8/c1-7-4-12(18)23-10-5-8(2-3-9(7)10)22-16-15(21)14(20)13(19)11(6-17)24-16/h2-5,11,13-17,19-21H,6H2,1H3/t11-,13-,14+,15-,16-/m1/s1

InChI key

YUDPTGPSBJVHCN-YMILTQATSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Methylumbelliferyl β-D-glucopyranoside has been used as substrate:
  • in glucosylceramidase β enzyme activity assay in lysosome-enriched fractions from primary hippocampal neurons
  • in β-glucosidase assay during yeast fermentation
  • to assay glucocerebrosidase 1 (GBA1)-related glucosidase activity in macrophage cell line (RAW)

Biochem/physiol Actions

4-Methylumbelliferyl β-D-glucopyranoside is a synthetic enzymatic substrate for glycosidase. It has been used as substrate for the β- glucosidase from enterococci.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An Improved Helferich Method for the alphabeta-Stereoselective Synthesis of 4-Methylumbelliferyl Glycosides for the Detection of Microorganisms
Wei X, et al.
Molecules (Basel), 20(12), 21681-21699 (2015)
Mitochondrial dysfunction and mitophagy defect triggered by heterozygous GBA mutations
Li H, et al.
Autophagy, 15(1), 113-130 (2019)
A Rashidi et al.
Bone marrow transplantation, 51(12), 1561-1564 (2016-08-16)
Post-transplant cyclophosphamide (PT-Cy) is the backbone of GvHD prophylaxis following haploidentical hematopoietic cell transplantation (haplo-HCT). PT-Cy has also been used in matched related (MRD) and unrelated (MUD) settings. It is not known whether outcomes are similar between haplo-HCT and MRD/MUD
Assessing microbial safety of drinking water improving approaches and methods: Improving approaches and methods, 253-253 (2003)
Florent Malard et al.
Biology of blood and marrow transplantation : journal of the American Society for Blood and Marrow Transplantation, 21(6), 1059-1067 (2015-02-25)
This retrospective report compared the 4-year outcomes of allogeneic stem cell transplantation (allo-SCT) in 651 adult patients with acute myeloid leukemia receiving a reduced-intensity (RIC) or nonmyeloablative conditioning (NMA) regimen according to the type of unrelated donors. These were either

Articles

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service