Skip to Content
Merck
All Photos(1)

Key Documents

I2771

Sigma-Aldrich

ITS+3 Liquid Media Supplement (100×)

liquid, sterile-filtered, BioReagent, suitable for cell culture

Synonym(s):

ITS Supplement

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352205
NACRES:
NA.75

sterility

sterile-filtered

product line

BioReagent

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

shipped in

ambient

storage temp.

2-8°C

General description

ITS+3 liquid media supplement is a mixture of insulin, human transferrin (partially iron-saturated), sodium selenite, linoleic acid, and bovine serum albumin (BSA). It is suitable for serum-free cell culture applications.

Application

ITS+3 Liquid Media Supplement (100×) has been used as a media supplement:
  • to support the growth of human adipose stem cells (ASC)
  • to culture porcine intervertebral disc (IVD) cells and tissues
  • to culture cartilage endplate (CEP) cells

Biochem/physiol Actions

  • Insulin: a polypeptide hormone that promotes glucose and amino acid uptake and may produce a mitogenic effect.
  • Transferrin: an iron-transport protein. Iron is an essential trace element but can be toxic in its free form. To nourish cells in culture, iron is supplied bound to transferrin in serum.
  • Selenium: an essential trace element present in serum.

Components

Contains 1.0 mg/ml bovine insulin, 0.55 mg/ml human transferrin (substantially iron-free), 0.5 μg/ml sodium selenite, 470 μg/ml linoleic acid, 470 μg/ml oleic acid and 50 mg/ml bovine serum albumin at the 100x concentration. Contains 2 moles each of linoleic acid and oleic acid per mole of albumin.

Preparation Note

Prepared in Earle′s Balanced Salt Solution (EBSS) without phenol red.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adam Cornish et al.
BMC genomics, 21(Suppl 11), 830-830 (2020-12-30)
Single-cell sequencing enables us to better understand genetic diseases, such as cancer or autoimmune disorders, which are often affected by changes in rare cells. Currently, no existing software is aimed at identifying single nucleotide variations or micro (1-50 bp) insertions and
Katherine Lakstins et al.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society, 39(6), 1204-1216 (2020-04-15)
Cartilage endplate degeneration/calcification has been linked to the onset and progression of intervertebral disc degeneration and there is a critical need to understand mechanisms, such as hypertrophic differentiation, of cartilage endplate degeneration/calcification to inform treatment strategies for discogenic back pain.
Xue Liu et al.
iScience, 24(6), 102551-102551 (2021-06-22)
Pulmonary mesenchymal cells are critical players in both the mouse and human during lung development and disease states. They are increasingly recognized as highly heterogeneous, but there is no consensus on subpopulations or discriminative markers for each subtype. We completed
Hien Lau et al.
PloS one, 15(12), e0243506-e0243506 (2020-12-08)
Previous studies have shown that necrostatin-1 (Nec-1) supplementation improved the viability of murine islets following exposure to nitric oxide, increased the survival of human islets during hypoxic culture, and augmented the maturation of pre-weaned porcine islets (PPIs) after 7 days
Di Wu et al.
International journal of stem cells, 14(1), 119-126 (2020-12-31)
The research on human hepatobiliary development and disorders has been constrained by minimal access to human fetal tissue, and low accuracy of animal models. To overcome this problem, we have established a system for the differentiation of human pluripotent stem

Articles

Importance and uses of linolenic acid in serum-free eukaryotic, including hybridoma and Chinese Hamster Ovary (CHO) cell, cultures

Understand the role of selenium in cell culture media formulations. The Media Expert library explains how culture media ingredients support cell growth.

Understand the role of selenium in cell culture media formulations. The Media Expert library explains how culture media ingredients support cell growth.

Understand the role of selenium in cell culture media formulations. The Media Expert library explains how culture media ingredients support cell growth.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service