Skip to Content
Merck
All Photos(1)

Key Documents

I1884

Sigma-Aldrich

Insulin-transferrin-sodium selenite media supplement

γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture

Synonym(s):

ITS Supplement

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352205
NACRES:
NA.75

biological source

human plasma

Quality Level

sterility

γ-irradiated

product line

BioXtra

form

lyophilized powder

technique(s)

cell culture | mammalian: suitable

UniProt accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

human ... INS(3630)

General description

Insulin-transferrin-sodium selenite media supplement supports in vitro growth and maturation of follicles from oocyte and is routinely used in in vitro fertilization methods.
Serum-free and reduced serum formulations often require purified forms of the factors normally supplied by serum that are suitable for in vitro growth and differentiation studies. These important factors include insulin, transferrin, selenium, pyruvate, and ethanolamine.

INSULIN is a polypeptide hormone that promotes the uptake of glucose and amino acids and may owe observed mitogenic effect to this property.

TRANSFERRIN is an iron-transport protein. Iron is an essential trace element, but can be toxic in the free form. To nourish cells in culture, it is supplied bound to transferrin in serum.

SELENIUM is an essential trace element normally provided by Serum.

Nutritional studies indicate that the supplement components described are utilized by most mammalian cells. They enhance cell proliferation and decrease the serum requirement of many cell types. When the provided supplement is used with 2 to 4 percent serum, proliferation is reported to be similar to medium supplemented with 10 percent serum.

Application

Insulin-transferrin-sodium selenite media supplement has been used as:
  • medium 199 (M199) media supplement for cardiac myocytes culture
  • Roswell Park Memorial Institute (RPMI) 1640 medium component for pluripotent stem cell (iPSC) culture
  • Dulbecco′s Modified Eagle Medium (DMEM) supplement in ID8 ovarian carcinoma cells

Other Notes

Each vial contains: ≥21 mg recombinant human insulin; ≥19 mg human transferrin (substantially iron free), and 25 μg sodium selenite.
ITS is a mixture of recombinant human insulin, human transferrin, and sodium selenite. It is a general cell supplement designed for use in non-complex media (e.g. MEM, RPMI-1640) and complex media (e.g. Ham?s F-12, DME/F-12, MEM) with sodium pyruvate.

Quantity

Each vial sufficient to prepare 5 liters of medium.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

TnI Structural Interface with the N-Terminal Lobe of TnC as a Determinant of Cardiac Contractility
Vetter AD, et al.
Biophysical Journal, 114(7), 1646-1656 (2018)
The effects of different concentrations of sodium selenite on the in vitro maturation of preantral follicles in serum-free and serum supplemented media
Abedelahi A, et al.
Journal of Assisted Reproduction and Genetics, 25(9-10), 483-488 (2008)
Bu-Er Wang et al.
PloS one, 3(5), e2186-e2186 (2008-05-15)
Although Wnt signaling has been shown to be important for embryonic morphogenesis and cancer pathogenesis of several tissues, its role in prostatic development and tumorigenesis is not well understood. Here we show that Wnt signaling regulated prostatic epithelial branching morphogenesis
Y Zhang et al.
Animal : an international journal of animal bioscience, 1-9 (2018-10-04)
Research on the causes of sheep death in sea voyages from Australia to the Middle East is limited, in particular little is known about the influence of climatic factors. Mortality data from 417 shipments of sheep exported over an 11-year
Michelle L Asp et al.
PloS one, 8(10), e78768-e78768 (2013-11-10)
Tamoxifen (Tam), a selective estrogen receptor modulator, is in wide clinical use for the treatment and prevention of breast cancer. High Tam doses have been used for treatment of gliomas and cancers with multiple drug resistance, but long QT Syndrome

Articles

Understand how transferrin affects serum-free cell culture systems, crucial for biomanufacturing heterologous proteins like monoclonal antibodies.

Understand how transferrin affects serum-free cell culture systems, crucial for biomanufacturing heterologous proteins like monoclonal antibodies.

Understand how transferrin affects serum-free cell culture systems, crucial for biomanufacturing heterologous proteins like monoclonal antibodies.

Understand how transferrin affects serum-free cell culture systems, crucial for biomanufacturing heterologous proteins like monoclonal antibodies.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service