Skip to Content
Merck
All Photos(1)

Key Documents

O3008

Sigma-Aldrich

Oleic Acid-Albumin from bovine serum

liquid, sterile-filtered, BioReagent, suitable for cell culture

Synonym(s):

Oleic acid-BSA complex

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352211
NACRES:
NA.75

biological source

bovine

sterility

sterile-filtered

product line

BioReagent

form

liquid

composition

Oleic acid, 2 mol/mol albumin

concentration

100 mg/mL BSA in DPBS
90-120 mg/mL protein (biuret)

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

storage temp.

2-8°C

Application

Oleic Acid-Albumin from bovine serum has been used: as a component of the Dulbecco′s modified Eagle′s medium (DMEM) to study adipogenic differentiation in chicken embryonic fibroblast cell lines, to study its effects on benign prostate epithelial cells as a component of the cell culture medium to quantify triglycerides in human epithelial colorectal adenocarcinoma cells

Biochem/physiol Actions

Fatty Acid-Albumin - complexes are used as alternative sources of lipids in the development of serum-free media. Fatty acids bind to serum proteins in high proportions. Serum proteins may help in the release of beneficial fatty acids and bind those that are inhibitory. Oleic acid bound to bovine serum albumin (BSA) shows a positive effect on the growth of various cells.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yinfang Wang et al.
Journal of experimental & clinical cancer research : CR, 38(1), 147-147 (2019-04-07)
Ionizing radiation (IR) therapy is the standard first-line treatment for newly diagnosed patients with glioblastoma (GBM), the most common and malignant primary brain tumor. However, the effects of IR are limited due to the aberrant radioresistance of GBM. Transcriptome analysis
Sarah Guttmann et al.
PloS one, 15(3), e0230025-e0230025 (2020-03-11)
Intestinal cells control delivery of lipids to the body by adsorption, storage and secretion. Copper (Cu) is an important trace element and has been shown to modulate lipid metabolism. Mutation of the liver Cu exporter ATP7B is the cause of
Maddalena Parafati et al.
International journal of molecular sciences, 21(24) (2020-12-19)
Non-alcoholic fatty liver disease (NAFLD) has a large impact on global health. At the onset of disease, NAFLD is characterized by hepatic steatosis defined by the accumulation of triglycerides stored as lipid droplets. Developing therapeutics against NAFLD and progression to
David M Favara et al.
Metabolites, 9(12) (2019-11-30)
Adhesion G Protein-Coupled Receptor L4 (ADGRL4/ELTD1) is an endothelial cell adhesion G protein-coupled receptor (aGPCR) which regulates physiological and tumour angiogenesis, providing an attractive target for anti-cancer therapeutics. To date, ADGRL4/ELTD1's full role and mechanism of function within endothelial biology
Timothy E Thayer et al.
Scientific reports, 10(1), 9831-9831 (2020-06-21)
Non-alcoholic fatty liver disease (NAFLD) affects over 30% of adults in the United States. Bone morphogenetic protein (BMP) signaling is known to contribute to hepatic fibrosis, but the role of BMP signaling in the development of NAFLD is unclear. In

Articles

Supplement cell culture systems with fatty acids for biomanufacturing heterologous proteins like monoclonal antibodies.

Supplement cell culture systems with fatty acids for biomanufacturing heterologous proteins like monoclonal antibodies.

Supplement cell culture systems with fatty acids for biomanufacturing heterologous proteins like monoclonal antibodies.

Supplement cell culture systems with fatty acids for biomanufacturing heterologous proteins like monoclonal antibodies.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service