Skip to Content
Merck
All Photos(6)

Key Documents

HPA017759

Sigma-Aldrich

Anti-FBN1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Fibrillin-1 precursor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

HCVSGMGMGRGNPEPPVSGEMDDNSLSPEACYECKINGYPKRGRKRRSTNETDASNIEDQSETEANVSLASWDVEKTAIFAFNISHVSNKVRILELLPALTTLTNHNRYLIESGNEDGFFKINQKEGISYLHFTKKKPVAGTYSLQISS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FBN1(2200)

General description

Fibrillin 1 (FBN1) is a 350kDa glycoprotein and a major constituent of microfibrils. The gene encoding it is localized on human chromosome 15. FBN1, which is a 2871 amino acid protein, has seven transforming growth factor-β1 binding protein-like (TB) domains and epidermal growth factor-like (EGF) domains.

Immunogen

Fibrillin-1 precursor recombinant protein epitope signature tag (PrEST)

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

Fibrillin 1 (FBN1) is an extracellular regulator of transforming growth factor β-1 (TGF-β1). It is a cytokine which is involved in the transdifferentiation of fibroblasts into myofibroblasts and also regulates microfibril assembly and stability. Mutations in the gene encoding FBN1 are known to be associated with Marfan syndrome (MFS).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72695

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tianyu Han et al.
Autophagy, 14(12), 2035-2048 (2018-09-21)
Macroautophagy/autophagy is a multistep cellular process that sequesters cytoplasmic components for lysosomal degradation. BECN1/Beclin1 is a central protein that assembles cofactors for the formation of a BECN1-PIK3C3-PIK3R4 complex to trigger the autophagy protein cascade. Discovering the regulators of BECN1 is
Chrishan J A Ramachandra et al.
International journal of cardiology, 187, 585-591 (2015-04-12)
Marfan syndrome (MFS) is a genetic disorder that affects multiple organs. Mortality imposed by aortic aneurysm and dissections represent the most serious clinical manifestation of MFS. Progressive pathological aortic root enlargement as the result of degeneration of microfibril architecture and
Fengyun Wang et al.
Molecular vision, 21, 194-200 (2015-03-03)
To screen mutations in the fibrillin-1 (FBN1) gene in a Chinese family with autosomal dominant Marfan syndrome (MFS). Patients and unaffected family members were given ophthalmic, cardiovascular, and physical examinations with a 5-year follow-up. Genomic DNA was extracted from the
K Tynan et al.
Human molecular genetics, 2(11), 1813-1821 (1993-11-01)
Marfan syndrome (MFS) is an autosomal dominantly inherited connective tissue disorder characterized by cardiovascular, ocular and skeletal manifestations. Previously, mutations in the fibrillin-1 gene on chromosome 15 (FBN1) have been reported to cause MFS. We have now screened 44 probands
Nagako Yoshiba et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 63(6), 438-448 (2015-03-26)
Myofibroblasts and extracellular matrix are important components in wound healing. Alpha-smooth muscle actin (α-SMA) is a marker of myofibroblasts. Fibrillin-1 is a major constituent of microfibrils and an extracellular-regulator of TGF-β1, an important cytokine in the transdifferentiation of resident fibroblasts

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service