Skip to Content
Merck
All Photos(1)

Documents

SAB4500863

Sigma-Aldrich

Anti-Fibrillin-1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

FBN, M350 kda glycoprotein component extracellular microfibril, MASS, MFS1, fibrillin-1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 312 kDa

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
immunohistochemistry: 1:50-1:100

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FBN1(2200)

Related Categories

General description

Fibrillin-1 is a multidomain cysteine-rich glycoprotein that belongs to the fibrillin protein family. It is encoded by the FBN1 gene mapped to human chromosome 15q21. Fibrillin-1 structure includes 43 calcium-binding epidermal growth factor (cbEGF)–like domains and 78 cysteine-containing TB motifs. The protein is ubiquitously present and is a vital component of elastic fiber–associated microfibrils in connective tissues. Anti-Fibrillin-1 Antibody detects endogenous levels of total Fibrillin-1 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human Fibrillin-1.

Immunogen Range: 2811-2860

Application

Anti-Fibrillin-1 antibody produced in rabbit has been used in immunohistochemical staining (1:50).

Biochem/physiol Actions

Fibrillin-1 protein plays an important role in the fibrillogenesis in elastic tissues and non-elastic tissues. It acts as an anchoring fiber and induces Smad2 signaling by regulating the bioavailability of endogenous transforming growth factor β1 (TGF-β1). Mutation in the FBN1 gene leads to the development of heritable disorder Marfan syndrome. Mutation of this gene also causes thoracic aortic aneurysms in patients without Marfan syndrome.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genetic Basis of Aortic Disease
Hawaiian Entomological Society., 91-100 (2018)
D M Milewicz et al.
Circulation, 94(11), 2708-2711 (1996-12-01)
Mutations in the FBN1 gene are the cause of the Marfan syndrome, an autosomal dominant disorder with skeletal, ocular, and cardiovascular complications. Aneurysms or dissections of the ascending thoracic aorta are the major cardiovascular complications of the disorder. We tested
Chapter 105 - Heritable Diseases of Connective Tissue
Hawaiian Entomological Society. (2016)
H C Dietz et al.
Human molecular genetics, 4 Spec No, 1799-1809 (1995-01-01)
The extracellular microfibril, 10-14 nm in diameter, performs a number of functions, including serving as the scaffolding for deposition of tropoelastin to form elastic fibers. A variety of proteins compose the structure of microfibrils, the most prominent of which are
Laure Delhon et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 33(2), 2707-2718 (2018-10-12)
Mutations in the a disintegrin and metalloproteinase with thrombospondin motif-like 2 ( ADAMTSL2) gene are responsible for the autosomal recessive form of geleophysic dysplasia, which is characterized by short stature, short extremities, and skeletal abnormalities. However, the exact function of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service