Skip to Content
Merck
All Photos(7)

Key Documents

HPA027066

Sigma-Aldrich

Anti-Fibronectin Antibody

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, rabbit polyclonal

Synonym(s):

Anti-CIG, Anti-FINC, Anti-GFND2, Anti-LETS, Anti-MSF, Anti-fibronectin 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

Product Name

Anti-FN1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

HEEICTTNEGVMYRIGDQWDKQHDMGHMMRCTCVGNGRGEWTCIAYSQLRDQCIVDDITYNVNDTFHKRHEEGHMLNCTCFGQGRGRWKCDPVDQCQDSETGTFYQIGDSWEKYVHGVRYQCYCYGRGIG

UniProt accession no.

application(s)

research pathology

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FN1(2335)

General description

Fibronectin 1 (FN1) is a glycoprotein of the extracellular matrix. It is encoded by the gene mapped to human chromosome 2q35. FN1 is expressed in the plasma and at the cell surface. Fibronectin is mostly synthesized by fibroblasts and endothelial cells.

Immunogen

fibronectin 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry. The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collecation of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Anti-FN1 antibody produced in rabbit has been used in western blotting and immunohistochemical staining.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

Fibronectin 1 (FN1) plays a vital role in cell adhesion, growth, migration, wound healing, blood coagulation and metastasis. In addition, it also facilitates cell attachment and proliferation, control of cell cytoskeleton, morphology and differentiation. FN1 is also involved in extracellular matrix formation, hemostasis and thrombosis. Mutations in FN1 lead to glomerulopathy. Fibronectin production by normal human foreskin fibroblasts has been shown to improve two-fold after glucocorticoid treatment.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72592

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CD93 promotes integrin-β1 activation and fibronectin fibrillogenesis during tumor angiogenesis.
Lugano, Roberta, et al.
The Journal of Clinical Investigation, 128(8), 3280?3297-3280?3297 (2018)
Tumor exosome-mediated promotion of adhesion to mesothelial cells in gastric cancer cells.
Arita T, et al.
Oncotarget, 7(35), 56855?56863-56855?56863 (2016)
S E Pors et al.
Human reproduction (Oxford, England), 34(8), 1523-1535 (2019-07-10)
Can a reconstructed ovary using decellularized human ovarian tissue (DCT) support survival of pre-antral stage follicles? We have demonstrated an effective protocol for decellularization of human ovarian tissues and successful recellularization with isolated human ovarian cells and pre-antral follicles. Survivors
Tomohiro Arita et al.
Oncotarget, 7(35), 56855-56863 (2016-08-04)
Peritoneal metastasis consists of a highly complex series of steps, and the details of the underlying molecular mechanism remain largely unclear. In this study, the effects of tumor-derived exosomes (TEX) on the progression of gastric cancers were investigated in peritoneal
Iris Kamer et al.
Molecular cancer research : MCR, 18(6), 926-937 (2020-03-15)
Tumor-host interactions play a major role in malignancies' initiation and progression. We have reported in the past that tumor cells attenuate genotoxic stress-induced p53 activation in neighboring stromal cells. Herein, we aim to further elucidate cancer cells' impact on signaling

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service