Skip to Content
Merck
All Photos(3)

Key Documents

04-035

Sigma-Aldrich

Anti-PTEN Antibody, clone 6H2.1

clone 6H2.1, Upstate®, from mouse

Synonym(s):

Anti-PTEN antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6H2.1, monoclonal

species reactivity

rat, human, mouse

manufacturer/tradename

Upstate®

technique(s)

immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

General description

The protein and lipid phosphatase PTEN (phosphatase and tensin homolog deleted on chromosome ten) has been suggested to play a role in tumorgenesis. PTEN is a tumor suppressor whose loss permits constitutive signaling through the PI3K pathway and is sufficient to cause cancer. In normal cells, in conjunction with FOXO1A and PTEN function together to restrain cell growth. PTEN metabolizes the product of PI3K phosphorylation of PI(4,5)P2 following receptor activation, creating PI(3,4,5)P3. In cells lacking PTEN, there are elevated levels of PI(3,4,5)P3, which acts as a potent second messenger promoting oncogenesis. Restoration of PTEN function is considered a novel approach to cancer therapy. Overexpression of PTEN inhibits cell migration, and expression of antisense PTEN mRNA enhances migration.

Specificity

recognizes PTEN

Immunogen

C-terminal 100 amino acids of PTEN
Epitope: C-terminal region

Application

Anti-PTEN Antibody, clone 6H2.1 detects level of PTEN & has been published & validated for use in WB, IP, IH(P).
Research Category
Signaling
Research Sub Category
PI3K, Akt, & mTOR Signaling

Quality

routinely evaluated on HeLa cell lysate

Target description

55 kDa

Linkage

Replaces: 07-016

Physical form

Format: Purified
Protein G Purified
Purified mouse IgG in storage buffer containing 0.1M Tris-Glycine, 0.15M NaCl, pH 7.4, with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dimitri G Trembath et al.
Melanoma research, 31(3), 264-267 (2021-04-20)
PTEN and p16 frequently undergo (epi)genetic aberrations in melanoma resulting in decreased, or absent, protein levels. We investigated the prognostic significance of these tumor suppressor genes in melanoma brain metastases (MBMs). Immunohistochemical analysis was performed on archived tissue sections from
Janire Mingo et al.
NPJ precision oncology, 3, 11-11 (2019-04-18)
Anti-PTEN monoclonal antibodies (mAb) are arising as important tools for immunohistochemistry (IHC) and protein quantification routine analysis in clinical oncology. Although an effort has been made to document the reliability of tumor tissue section immunostaining by anti-PTEN mAb, and to
BRCAness profile of sporadic ovarian cancer predicts disease recurrence.
Wysham, WZ; Mhawech-Fauceglia, P; Li, H; Hays, L; Syriac, S; Skrepnik, T; Wright et al.
Testing null
Victoria K Hill et al.
PloS one, 12(5), e0176683-e0176683 (2017-05-04)
PTEN is among the most commonly mutated tumor suppressor genes in human cancer. However, studying the role of PTEN in the pathogenesis of cancer has been limited, in part, by the paucity of human cell-based isogenic systems that faithfully model
Whole chromosome instability caused by Bub1 insufficiency drives tumorigenesis through tumor suppressor gene loss of heterozygosity.
Baker, DJ; Jin, F; Jeganathan, KB; van Deursen, JM
Cancer Cell null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service