Skip to Content
Merck
All Photos(1)

Key Documents

80518

Sigma-Aldrich

Sodium thiocyanate solution

BioUltra, 8 M in H2O

Synonym(s):

Sodium rhodanide solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CNNaS
CAS Number:
Molecular Weight:
81.07
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.26
form:
liquid
solubility:
H2O: 8 M at 20 °C, clear, colorless

product line

BioUltra

Quality Level

form

liquid

concentration

8 M in H2O

impurities

insoluble matter, passes filter test

pH

5-8 (25 °C, 8 M in H2O)

solubility

H2O: 8 M at 20 °C, clear, colorless

density

1.295 g/mL at 20 °C

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤100 mg/kg
sulfide (S2-): ≤20 mg/kg

cation traces

Al: ≤1 mg/kg
As: ≤0.1 mg/kg
Ba: ≤1 mg/kg
Bi: ≤1 mg/kg
Ca: ≤50 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
K: ≤200 mg/kg
Li: ≤1 mg/kg
Mg: ≤1 mg/kg
Mn: ≤1 mg/kg
Mo: ≤1 mg/kg
NH4+: ≤40 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Sr: ≤1 mg/kg
Zn: ≤1 mg/kg

absorption

cut-off at 276 nm in H2O at 8 M

SMILES string

[Na+].[N-]=C=S

InChI

1S/CHNS.Na/c2-1-3;/h3H;/q;+1/p-1

InChI key

VGTPCRGMBIAPIM-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application


  • Investigation of supramolecular structures in various aqueous solutions of an amyloid forming peptide using small-angle X-ray scattering.: Sodium thiocyanate solution was utilized to investigate the supramolecular structures of amyloid-forming peptides, providing insights into amyloid diseases and potential therapeutic approaches (Brunzell et al., 2024).

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sandra Romero-Steiner et al.
Clinical and diagnostic laboratory immunology, 12(9), 1029-1035 (2005-09-09)
Determination of antibody avidity measurements can be difficult in human serum depending on the population evaluated. We evaluated three approaches for the determination of antibody avidity for immunoglobulin G (IgG). These approaches were (i) elution of bound antibody with increasing
Vincent Ball et al.
Langmuir : the ACS journal of surfaces and colloids, 21(9), 4129-4137 (2005-04-20)
In this study, we investigate the buildup of PEI-(PSS-PAH)(n) polyelectrolyte multilayers at pH 7.4 in the presence of either NaCl or NaSCN as a supporting electrolyte. It appears that in the presence of increasing thiocyanate concentrations (from 0.1 to 0.5
Barbaros Ozer et al.
The Journal of dairy research, 70(2), 227-232 (2003-06-13)
The effects of activation of the lactoperoxidase (LPO) system by H2O2-NaSCN and hydrogen peroxide (H2O2) on the accessibility of sulphydryl groups (SH) in skimmed milk, and on the dynamic rheological properties of the resulting yoghurt were investigated. Four different concentrations
Maurits C R Heijna et al.
The journal of physical chemistry. B, 111(7), 1567-1573 (2007-02-01)
In protein crystallography, spherulites are considered the result of a failed crystallization experiment. Understanding the formation of these structures may contribute to finding methods to prevent their formation. Here, we present an in situ study on lysozyme spherulites grown from
A S Taisova et al.
Biofizika, 49(6), 1069-1074 (2004-12-23)
The properties of the light-harvesting superantenna of the photosynthesizing bacteria from the new family of green filamentous bacteria Oscillochloridaceae were investigated by optical spectroscopy. The antenna of Oscillochloris trichoides consists of peripheral chlorosomal and membrane subantennas. A method of isolation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service