Skip to Content
Merck
All Photos(2)

Key Documents

O652

Sigma-Aldrich

Octadecane

99%

Synonym(s):

C18-n-Alkane, n-Octadecane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)16CH3
CAS Number:
Molecular Weight:
254.49
Beilstein:
1740138
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

8.8 (vs air)

Quality Level

vapor pressure

1 mmHg ( 119 °C)

Assay

99%

form

solid or liquid (Conforms to requirements)

autoignition temp.

455 °F

bp

317 °C (lit.)

mp

26-29 °C (lit.)

density

0.777 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCC

InChI

1S/C18H38/c1-3-5-7-9-11-13-15-17-18-16-14-12-10-8-6-4-2/h3-18H2,1-2H3

InChI key

RZJRJXONCZWCBN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

n-Octadecane is generally used in developing microencapsulated phase-change materials.
It can also be used as a reactant to synthesize:
  • N-octadecylsulfonyl)-alanine sodium salt
  • 1-octadecanesulfonyl chloride
  • 2-octadecanone

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

330.8 °F - closed cup

Flash Point(C)

166 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aiym Tleuova et al.
Nanomaterials (Basel, Switzerland), 10(12) (2020-12-03)
Pyraclostrobin (Pyr) is one of the most effective fungicides. However, it can degrade via photolysis in water, it is toxic to aquatic life and if inhaled, it has a low solubility in water, that leads to difficulties when applying to
Synthesis and Physicochemical Properties of Alanine?Based Surfactants.
Bougueroua M, et al.
Journal of Surfactants and Detergents, 19(6), 1121-1131 (2016)
Ahmad Fariz Nicholas et al.
Nanomaterials (Basel, Switzerland), 8(9) (2018-09-08)
The preparation of activated carbon using palm kernel shells as the precursor (PKSAC) was successfully accomplished after the parametric optimization of the carbonization temperature, carbonization holding time, and the ratio of the activator (H₃PO₄) to the precursor. Optimization at 500
Fundamental time scales governing organic aerosol multiphase partitioning and oxidative aging.
Zhang H, et al.
Environmental Science & Technology, 49(16), 9768-9777 (2015)
Fabrication and performances of microencapsulated phase change materials based on n-octadecane core and resorcinol-modified melamine?formaldehyde shell.
Zhang H and Wang X
Colloids Surf., 332(2-3), 129-138 (2009)

Protocols

Separation of Decane; Dodecane; Tetradecane; Hexadecane; Octadecane; Eicosane; Docosane; Tetracosane; Hexacosane; Octacosane

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service