Skip to Content
Merck
All Photos(1)

Key Documents

52209

Supelco

Hexadecane

analytical standard

Synonym(s):

n-Hexadecane, Cetane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)14CH3
CAS Number:
Molecular Weight:
226.44
Beilstein:
1736592
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

7.8 (vs air)

vapor pressure

1 mmHg ( 105.3 °C)

Assay

≥99.8% (GC)

autoignition temp.

395 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.434 (lit.)
n20/D 1.435

bp

287 °C (lit.)

mp

18 °C (lit.)

transition temp

solidification point 17.5-18.5 °C

density

0.773 g/mL at 25 °C (lit.)

application(s)

petroleum

format

neat

SMILES string

CCCCCCCCCCCCCCCC

InChI

1S/C16H34/c1-3-5-7-9-11-13-15-16-14-12-10-8-6-4-2/h3-16H2,1-2H3

InChI key

DCAYPVUWAIABOU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hexadecane can find applications as a sole carbon source for the growth of canals, which are supramolecular structures observed in the cell wall of Candida maltose.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

233.6 °F

Flash Point(C)

112 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Cell wall canals formed upon growth of Candida maltosa in the presence of hexadecane are associated with polyphosphates
Zvonarev, NA
FEMS Yeast Research, 17 (2017)
Shweta Mishra et al.
Bioresource technology, 111, 148-154 (2012-03-13)
In the present study, n-hexadecane degradation in MSM was investigated by three bacteria identified as Pseudomonas aeruginosa PSA5, Rhodococcus sp. NJ2 and Ochrobactrum intermedium P2, isolated from petroleum sludge. During 10 days of incubation, n-hexadecane was degraded to 99% by
Feliciano Priego-Capote et al.
Analytical chemistry, 80(8), 2881-2887 (2008-03-14)
A new approach based on miniemulsion polymerization is demonstrated for synthesis of molecularly imprinted nanoparticles (MIP-NP; 30-150 nm) with "monoclonal" binding behavior. The performance of the MIP nanoparticles is characterized with partial filling capillary electrochromatography, for the analysis of rac-propranolol
Diana Gutiérrez et al.
Applied and environmental microbiology, 81(10), 3336-3348 (2015-03-10)
Phage therapy is a promising option for fighting against staphylococcal infections. Two lytic phages, vB_SauM_phiIPLA-RODI (phiIPLA-RODI) and vB_SepM_phiIPLA-C1C (phiIPLA-C1C), belonging to the Myoviridae family and exhibiting wide host ranges, were characterized in this study. The complete genome sequences comprised 142,348
Matthew A Churchward et al.
Biophysical journal, 94(10), 3976-3986 (2008-01-30)
The Ca(2+)-triggered merger of two apposed membranes is the defining step of regulated exocytosis. CHOL is required at critical levels in secretory vesicle membranes to enable efficient, native membrane fusion: CHOL-sphingomyelin enriched microdomains organize the site and regulate fusion efficiency

Protocols

Separation of Decane; Dodecane; Tetradecane; Hexadecane; Octadecane; Eicosane; Docosane; Tetracosane; Hexacosane; Octacosane

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service