Skip to Content
Merck
All Photos(1)

Key Documents

B0300

Sigma-Aldrich

Betaine solution

5 M, PCR Reagent

Synonym(s):

TMG, trimethylglycine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3N+CH2COO
CAS Number:
Molecular Weight:
117.15
Beilstein:
3537113
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.52

grade

PCR Reagent

Quality Level

form

liquid

packaging

vial of 1.5 mL

concentration

5 M

technique(s)

PCR: suitable

color

colorless

application(s)

agriculture

foreign activity

DNase, RNase, none detected

storage temp.

2-8°C

SMILES string

C[N+](C)(C)CC([O-])=O

InChI

1S/C5H11NO2/c1-6(2,3)4-5(7)8/h4H2,1-3H3

InChI key

KWIUHFFTVRNATP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Betaine also called trimethylglycine or N,N,N triethylammonium acetate, is an analog of glycine with three methyl groups. It is highly compatible with polymerase chain reaction (PCR) buffer mixture. Betaine is a PCR enhancing reagent that is widely used for improving the yield and specificity of PCR products, especially during the PCR amplification of targets rich in GC content or those that form secondary structures resulting in poor yield. Betaine facilitates DNA strand separation and manages the DNA melting temperature (Tm) difference between the GC and AT pairs in DNA. It stabilizes the ds DNA by equalizing the contribution of GC- and AT-base pairs. Betaine has been broadly used to optimize multiplex and ‘long and accurate′ polymerase chain reaction (LA-PCR). The addition of 1.0-1.7 M aqueous betaine to a PCR mixture has been reported to reduce the base pair composition dependence on DNA strand melting.

Application

Betaine solution has been used as a component of the reaction mixture for:
  • loop-mediated isothermal amplification (LAMP) reactions
  • polymerase chain reaction for genomic DNA amplification
  • quantitative polymerase chain reaction (qPCR) for telomere length analysis
  • in the PCR amplification of CGG repeats in the genomic DNA
  • reverse-transcription loop-mediated isothermal amplification (RT-LAMP) to detect severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2)
  • PCR amplification of DNA-directed RNA polymerase II largest subunit (RPB2)
  • reverse transcription during library preparation using Smartseq2

Packaging

1.5mL per vial

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thanyaornwanya Charoenwijitkul et al.
Clinical biochemistry, 71, 31-37 (2019-06-15)
The 3.7 kb deletion (-α3.7) is the most common form of α+-thalassemia found in multiple populations which can be classified into three subtypes. In order not to mis-identify it, the molecular information within each population is required. We have addressed this
P N Hengen
Trends in biochemical sciences, 22(6), 225-226 (1997-06-01)
Methods and reagents is a unique monthly column that highlights current discussions in the newsgroup bionet.molbio.methds-reagnts, available on the internet. This month's column discusses the use of additives for optimizing the amount and quality of product obtained through multiplex and
Betaine chemistry, roles, and potential use in liver disease
Day CR and Kempson SA
Biochimica et biophysica acta. General subjects, 1860(6), 1098-1106 (2016)
Secondary metabolites of Phlebopus species from Northern Thailand
Chuankid B, et al.
Mycological research, 19(12), 1525-1536 (2020)
Thirty-minute screening of antibiotic resistance genes in bacterial isolates with minimal sample preparation in static self-dispensing 64 and 384 assay cards
Kostic T, et al.
Applied Microbiology and Biotechnology, 99(18), 7711-7722 (2015)

Articles

SeqPlex™-I WTA kit amplifies RNA for NGS, enabling genomic studies from limited samples.

SeqPlex™-I WTA kit amplifies RNA for NGS, enabling genomic studies from limited samples.

SeqPlex™-I WTA kit amplifies RNA for NGS, enabling genomic studies from limited samples.

SeqPlex™-I WTA kit amplifies RNA for NGS, enabling genomic studies from limited samples.

Protocols

Protocol for high fidelity amplification of long PCR fragments up to 22kb from complex DNA mixtures and up to 40kb from simple DNA mixtures. AccuTaq LA.

REDAccuTaq LA protocol offers high-fidelity amplification of long PCR fragments with direct gel loading capability.

REDAccuTaq LA protocol offers high-fidelity amplification of long PCR fragments with direct gel loading capability.

REDAccuTaq LA protocol offers high-fidelity amplification of long PCR fragments with direct gel loading capability.

See All

Related Content

KOD One™ PCR Master Mix overview for ultra-fast PCR with high specificity, fidelity, and yield

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service