Skip to Content
Merck
All Photos(1)

Key Documents

46960

Sigma-Aldrich

Fluorescein sodium salt

BioReagent, suitable for fluorescence

Synonym(s):

Acid Yellow 73, NaFl, Sodium fluorescein, Yellow No. 8, D&C;Yellow No. 8, NaFluo, Uranine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H10Na2O5
CAS Number:
Molecular Weight:
376.27
Colour Index Number:
45350
Beilstein:
3833041
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.32

product line

BioReagent

form

powder

technique(s)

titration: suitable

fluorescence

λex 460 nm; λem 515 nm(lit.)
λex 490 nm; λem 514 nm in 0.1 M Tris pH 8.0

suitability

suitable for fluorescence

SMILES string

[Na+].[Na+].[O-]c1ccc2c(Oc3cc([O-])ccc3C24OC(=O)c5ccccc45)c1

InChI

1S/C20H12O5.2Na/c21-11-5-7-15-17(9-11)24-18-10-12(22)6-8-16(18)20(15)14-4-2-1-3-13(14)19(23)25-20;;/h1-10,21-22H;;/q;2*+1/p-2

InChI key

RGPLGPBQJOQFJS-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Fluorescein Sodium Salt (FSS) is a xanthene dye having a polycrystalline structure with a monoclinic system. Fluorescein is a synthetic organic compound available as a dark orange/red powder soluble in water and alcohol.

Application

Fluorescein Sodium Salt (FSS) is used to conduct in-vivo skin studies.

Xanthene fluorophores, like Fluorescein Sodium Salt (FSS), are versatile compounds used across diverse fields of chemistry and life sciences.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

423.7 °F - Pensky-Martens closed cup

Flash Point(C)

217.6 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joshua L Turnbull et al.
Journal of the American Chemical Society, 143(16), 6194-6201 (2021-04-03)
Xanthene fluorophores, like fluorescein, have been versatile molecules across diverse fields of chemistry and life sciences. Despite the ubiquity of 3-carboxy and 3-sulfonofluorescein for the last 150 years, to date, no reports of 3-phosphonofluorescein exist. Here, we report the synthesis
Christopher Price et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 26(2), 277-285 (2010-08-18)
Since proposed by Piekarski and Munro in 1977, load-induced fluid flow through the bone lacunar-canalicular system (LCS) has been accepted as critical for bone metabolism, mechanotransduction, and adaptation. However, direct unequivocal observation and quantification of load-induced fluid and solute convection
Sara Seabrooke et al.
American journal of physiology. Cell physiology, 305(5), C558-C567 (2013-06-28)
The blood-brain barrier (BBB) physiologically isolates the brain from the blood and, thus, plays a vital role in brain homeostasis. Ion transporters play a critical role in this process by effectively regulating access of chemicals to the brain. Organic anion-transporting
Tomoaki Murakami et al.
Ophthalmology, 120(12), 2596-2603 (2013-08-21)
To study the association between the fluorescence levels on fluorescein angiography images and the characteristics on spectral-domain optical coherence tomography (SD OCT) images in diabetic macular edema (DME). Retrospective, observational, cross-sectional study. One hundred sixty-seven consecutive eyes of 116 patients
Wee Yang Ng et al.
Biomicrofluidics, 3(2), 22405-22405 (2009-08-21)
This paper investigates the phenomenon of Faradaic charging in ac electrokinetics. Faradaic reactions were suggested as a key effect responsible for the reversal of pumping direction in ac micropumps. However, this hypothesis has yet to be proven convincingly and directly.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service