D87759
7-Diethylamino-4-methylcoumarin
99%
Synonym(s):
Coumarin 1
Sign Into View Organizational & Contract Pricing
All Photos(4)
About This Item
Empirical Formula (Hill Notation):
C14H17NO2
CAS Number:
Molecular Weight:
231.29
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Quality Level
Assay
99%
form
solid
mp
72-75 °C (lit.)
SMILES string
CCN(CC)c1ccc2C(C)=CC(=O)Oc2c1
InChI
1S/C14H17NO2/c1-4-15(5-2)11-6-7-12-10(3)8-14(16)17-13(12)9-11/h6-9H,4-5H2,1-3H3
InChI key
AFYCEAFSNDLKSX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
7-Diethylamino-4-methylcoumarin (C1) is a bichromophoric fluorescent laser dye, which has a laser action at 460 nm.
Application
Terbium based metal organic framework may be doped with C1 for potential use in ratiometric temperature sensing. C1 may also be used as a fluorescent photo-initiator that can act as an optical brightener for use in the synthesis of the light emitting diode (LED) based hydrogel.
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Novel applications of fluorescent brighteners in aqueous visible-light photopolymerization: high performance water-based coating and LED-assisted hydrogel synthesis
Zuo X, et al.
Polym. Chem., 9(28), 3952-3958 (2018)
Fluorescent tracer and pesticide penetration through selected protective clothing.
B A Archibald et al.
Bulletin of environmental contamination and toxicology, 53(4), 479-485 (1994-10-01)
Synthesis and spectroscopic characterization of coumarin 440 dye doped silica gel rods
Haque FZ and Husain M
Optik, 125(10), 2308-2312 (2014)
R K Oestenstad et al.
American Industrial Hygiene Association journal, 51(5), 280-284 (1990-05-01)
A method has been developed to identify the location and shape of respirator faceseal leak sites by the deposition of a fluorescent tracer. An aerosol generation, conditioning, and exposure system to provide a test environment with stable aerosol concentration and
A dye encapsulated terbium-based metal-organic framework for ratiometric temperature sensing
Xia T, et al.
Dalton Transactions, 45(46), 18689-18695 (2016)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service