Skip to Content
Merck
All Photos(3)

Key Documents

F6377

Sigma-Aldrich

Fluorescein sodium salt

Powder

Synonym(s):

Acid Yellow 73, D&C;Yellow No. 8, NaFl, NaFluo, Sodium fluorescein, Uranine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H10Na2O5
CAS Number:
Molecular Weight:
376.27
Colour Index Number:
45350
Beilstein:
3833041
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

Fluorescein sodium salt, used as fluorescent tracer

Quality Level

form

powder

technique(s)

titration: suitable

pH

8.3 (20 °C, 10 g/L)

mp

>300 °C

solubility

H2O: 1 mg/mL

density

0.78 g/cm3 at 29.6 °C (85.3 °F at 699.7 hPa)

fluorescence

λex 460 nm; λem 515 nm(lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].[O-]c1ccc2c(Oc3cc([O-])ccc3C24OC(=O)c5ccccc45)c1

InChI

1S/C20H12O5.2Na/c21-11-5-7-15-17(9-11)24-18-10-12(22)6-8-16(18)20(15)14-4-2-1-3-13(14)19(23)25-20;;/h1-10,21-22H;;/q;2*+1/p-2

InChI key

RGPLGPBQJOQFJS-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Fluorescein is a xanthene dye and is a common labeling agent in biology.

Application

Fluorescein sodium salt has been used as a solute to study microvessel permeability. It has been used as a test molecule to study blood barrier penetration.
Water soluble fluorescent dye used in permeability and vascular perfusion studies due to its small molecular radius. Sodium fluorescein is used as a fluorescent tracer to study blood-brain barrier (BBB) and blood-spinal cord barrier (BSCB) permeability in rodent models. The dye was used as a probe substrate in the study of organic anion transporting polypeptide (OATP) mediated drug transport in hepatocytes. Sodium fluorescein was also used to monitor in situ solute transportation into bone through fluorescence recovery after photobleaching (FRAP).

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

423.7 °F - Pensky-Martens closed cup

Flash Point(C)

217.6 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Montermini et al.
The Journal of physiology, 543(Pt 3), 959-975 (2002-09-17)
The permeability, P(S), to sodium fluorescein (Stokes-Einstein radius = 0.45 nm) has been measured in single mesenteric capillaries of pithed frogs and anaesthetised rats as perfusion velocity, U, was varied over a range from 400 up to 2000-10,000 microm s(-1).
P Kozler et al.
Physiological research, 52(5), 607-614 (2003-10-11)
The aim was to study the blood-brain permeability according to the distribution in the rat brain of Evans blue (EB) and sodium fluorescein (NaFl) administered by an intracarotid injection. Eighteen animals were divided into six groups according to the state
Temperature dependence of electronic parameters of organic Schottky diode based on fluorescein sodium salt.
Yahia IS, et al.
Synthetic Metals, 161(9), 881-887 (2011)
The acute and chronic toxicity of two xanthene dyes, fluorescein sodium salt and phloxine B, to Daphnia pulex.
Walthall WK and Stark JD.
Environmental Pollution (Barking, Essex : 1987), 104(2), 207-215 (1999)
Vascular endothelial growth factor enhances cancer cell adhesion to microvascular endothelium in vivo.
Shen S
Experimental Physiology, 95, 369-369 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service