Skip to Content
Merck
All Photos(1)

Key Documents

46970

Sigma-Aldrich

Fluorescein sodium salt

p.a., for source staining

Synonym(s):

Acid Yellow 73, D&C;Yellow No. 8, NaFl, NaFluo, Sodium fluorescein, Uranine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H10Na2O5
CAS Number:
Molecular Weight:
376.27
Colour Index Number:
45350
Beilstein:
3833041
EC Number:
MDL number:
UNSPSC Code:
12171501
PubChem Substance ID:
NACRES:
NA.32

grade

for source staining
p.a.

form

powder

fluorescence

λex 460 nm; λem 515 nm(lit.)

SMILES string

[Na+].[Na+].[O-]c1ccc2c(Oc3cc([O-])ccc3C24OC(=O)c5ccccc45)c1

InChI

1S/C20H12O5.2Na/c21-11-5-7-15-17(9-11)24-18-10-12(22)6-8-16(18)20(15)14-4-2-1-3-13(14)19(23)25-20;;/h1-10,21-22H;;/q;2*+1/p-2

InChI key

RGPLGPBQJOQFJS-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Fluorescein Sodium Salt (FSS) is a xanthene dye having a polycrystalline structure with a monoclinic system. Fluorescein is a synthetic organic compound available as a dark orange/red powder soluble in water and alcohol.

Application

Fluorescein Sodium Salt (FSS) is used to conduct in-vivo skin studies.

Xanthene fluorophores, like Fluorescein Sodium Salt (FSS), are versatile compounds used across diverse fields of chemistry and life sciences.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

423.7 °F - Pensky-Martens closed cup

Flash Point(C)

217.6 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jacob Beal et al.
Communications biology, 3(1), 512-512 (2020-09-19)
Optical density (OD) is widely used to estimate the density of cells in liquid culture, but cannot be compared between instruments without a standardized calibration protocol and is challenging to relate to actual cell count. We address this with an
Joshua L Turnbull et al.
Journal of the American Chemical Society, 143(16), 6194-6201 (2021-04-03)
Xanthene fluorophores, like fluorescein, have been versatile molecules across diverse fields of chemistry and life sciences. Despite the ubiquity of 3-carboxy and 3-sulfonofluorescein for the last 150 years, to date, no reports of 3-phosphonofluorescein exist. Here, we report the synthesis
Beau J Fenner et al.
Investigative ophthalmology & visual science, 54(13), 8013-8019 (2013-11-14)
To determine the reliability of single- and double-zone corneal fluorescent staining compared with five-zone analysis for the prediction of dry eye disease. Prospective study of 510 subjects with dry eye disease characterized using corneal fluorescein staining, Schirmer scores, and tear
Tomoaki Murakami et al.
Ophthalmology, 120(12), 2596-2603 (2013-08-21)
To study the association between the fluorescence levels on fluorescein angiography images and the characteristics on spectral-domain optical coherence tomography (SD OCT) images in diabetic macular edema (DME). Retrospective, observational, cross-sectional study. One hundred sixty-seven consecutive eyes of 116 patients
Christopher Price et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 26(2), 277-285 (2010-08-18)
Since proposed by Piekarski and Munro in 1977, load-induced fluid flow through the bone lacunar-canalicular system (LCS) has been accepted as critical for bone metabolism, mechanotransduction, and adaptation. However, direct unequivocal observation and quantification of load-induced fluid and solute convection

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service