Skip to Content
Merck
All Photos(2)

Key Documents

414220

Sigma-Aldrich

Propylene carbonate

suitable for HPLC, 99.7%

Synonym(s):

1,2-Propanediol cyclic carbonate, 4-Methyl-1,3-dioxolan-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6O3
CAS Number:
Molecular Weight:
102.09
Beilstein:
107913
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
NACRES:
NA.21

vapor pressure

0.13 mmHg ( 20 °C)
0.98 mmHg ( 50 °C)

Assay

99.7%

form

liquid

autoignition temp.

851 °F

expl. lim.

14.3 %

technique(s)

HPLC: suitable

impurities

<0.020% water

evapn. residue

<0.0005%

refractive index

n20/D 1.421 (lit.)

pH

7 (20 °C, 200 g/L)

bp

240 °C (lit.)

mp

−55 °C (lit.)

density

1.204 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 235 nm Amax: 1.00
λ: 280 nm Amax: 0.50
λ: 300 nm Amax: 0.30
λ: 350 nm Amax: 0.05
λ: 375-400 nm Amax: 0.01

application(s)

food and beverages

SMILES string

CC1COC(=O)O1

InChI

1S/C4H6O3/c1-3-2-6-4(5)7-3/h3H,2H2,1H3

InChI key

RUOJZAUFBMNUDX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Propylene carbonate is a cyclic carbonate commonly used as an organic solvent. Many methods have been reported for its synthesis from propylene oxide and carbon dioxide under different reaction conditions. The dielectric property, relaxation times and viscosity has been studied. It is a potential electrochemical solvent in preparing aprotic battery electrolytes.
This solvent is glass distilled, submicron filtered and subjected to rigorous specification testing to provide lot-to-lot consistency.

Application

Designed for use with HPLC instrumentation and organic synthesis applications.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

269.6 °F - closed cup

Flash Point(C)

132 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New procedure for recycling homogeneous catalyst: propylene carbonate synthesis under supercritical CO2 conditions.
He LN, et al.
Green Chemistry, 5(1), 92-94 (2003)
Exfoliation of graphite oxide in propylene carbonate and thermal reduction of the resulting graphene oxide platelets.
Zhu Y, et al.
ACS Nano, 4(2), 1227-1233 (2010)
Kuhnel I, et al.
Holzforschung, 69(5), 531-538 (2015)
Synthesis of finely divided molybdenum sulfide nanoparticles in propylene carbonate solution.
Afanasiev P.
Journal of Solid State Chemistry, 213, 158-164 (2014)
A rapid and effective synthesis of propylene carbonate using a supercritical CO2-ionic liquid system.
Kawanami H, et al.
Chemical Communications (Cambridge, England), 7, 896-897 (2003)

Articles

Solid-state lithium fast-ion conductors are crucial for safer, high-energy-density all-solid-state batteries, addressing conventional battery limitations.

Solid-state lithium fast-ion conductors are crucial for safer, high-energy-density all-solid-state batteries, addressing conventional battery limitations.

Solid-state lithium fast-ion conductors are crucial for safer, high-energy-density all-solid-state batteries, addressing conventional battery limitations.

Solid-state lithium fast-ion conductors are crucial for safer, high-energy-density all-solid-state batteries, addressing conventional battery limitations.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service