Skip to Content
Merck
All Photos(1)

Key Documents

494496

Sigma-Aldrich

NMP

biotech. grade, ≥99.7%

Synonym(s):

1-Methyl-2-pyrrolidone, N-Methyl-2-pyrrolidone, NMP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO
CAS Number:
Molecular Weight:
99.13
Beilstein:
106420
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.07

product name

1-Methyl-2-pyrrolidinone, biotech. grade, ≥99.7%

grade

biotech. grade

Quality Level

vapor density

3.4 (vs air)

vapor pressure

0.29 mmHg ( 20 °C)
0.99 mmHg ( 40 °C)

Assay

≥99.7%

form

liquid

autoignition temp.

518 °F

expl. lim.

9.5 %

impurities

≤0.005% water
≤0.01% free amines (CH3NH2)

color

APHA: ≤20

refractive index

n20/D 1.47 (lit.)

pH

7.7-10.0 (20 °C, 100 g/L)

bp

202 °C (lit.)
81-82 °C/10 mmHg (lit.)

mp

−24 °C (lit.)

solubility

acetone: miscible(lit.)
alcohol: miscible(lit.)
chloroform: miscible(lit.)
ethyl acetate: miscible(lit.)
water: miscible(lit.)

density

1.028 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 285 nm Amax: 1.00
λ: 300 nm Amax: 0.50
λ: 325 nm Amax: 0.10
λ: 350-400 nm Amax: 0.01

application(s)

peptide synthesis

SMILES string

CN1CCCC1=O

InChI

1S/C5H9NO/c1-6-4-2-3-5(6)7/h2-4H2,1H3

InChI key

SECXISVLQFMRJM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N-Methyl-2-pyrrolidinone (N-methylpyrrolidone, NMP) is a polar cyclic organic solvent. A study on the impact of NMP on Saccharomyces cerevisiae revealed that it induced aneuploidy. Characteristic properties of NMP include low viscosity, excellent dissolving power and enhanced demulsifying property. It is a better alternative to phenol in selectively treating lube feedstock. The behavior of fullerene-C60 in NMP solution has been examined based on electronic absorption and photoluminescence spectral data. A solution of sodium borohydride (NaBH4) in NMP showed increased reactivity towards debromination of alkyl bromide. The enhanced selectivity of NMP-ethylene glycol solvent mixture for aromatic hydrocarbon extraction has been analyzed by gas chromatography.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

195.8 °F - Pensky-Martens closed cup

Flash Point(C)

91 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

NaBH4 in N-methylpyrrolidone: a safe alternative for hydride displacement reaction.
Torisawa Y, et al.
Bioorganic & Medicinal Chemistry Letters, 11(20), 2787-2789 (2001)
Evaluation of extraction properties of N-methylpyrrolidone and ethylene glycol mixtures by gas chromatography.
Wardencki W and Tameesh AHH.
Journal of Chemical Technology and Biotechnology, 31(1), 86-92 (1981)
Aggregation of fullerene C60 in N-methylpyrrolidone.
Yevlampieva NP, et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 209(2), 167-171 (2002)
Sanjairaj Vijayavenkataraman et al.
Artificial organs, 43(5), 515-523 (2018-09-20)
The incidence of peripheral nerve injuries is on the rise and the current gold standard for treatment of such injuries is nerve autografting. Given the severe limitations of nerve autografts which include donor site morbidity and limited supply, neural guide
N-methylpyrrolidone instead of phenol in treatment of lube feedstock.
Ivanov AV, et al.
Chemistry and Technology of Fuels and Oils, 36(5), 352-354 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service