Skip to Content
Merck
All Photos(3)

Key Documents

431567

Sigma-Aldrich

Lithium perchlorate

greener alternative

99.99% trace metals basis

Synonym(s):

Perchloric acid, lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiClO4
CAS Number:
Molecular Weight:
106.39
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.99% trace metals basis
grade:
ACS reagent
form:
granular
solubility:
H2O: 106.4 g/L at 20 °C

grade

ACS reagent

Quality Level

Assay

99.99% trace metals basis

form

granular

reaction suitability

reagent type: oxidant

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤0.005% insolubles
<100 ppm total metallic impurities

pH

6.0-7.5

mp

236 °C (lit.)

solubility

H2O: 106.4 g/L at 20 °C

anion traces

chloride (Cl-): ≤0.003%
sulfate (SO42-): ≤0.001%

greener alternative category

SMILES string

[Li+].[O-]Cl(=O)(=O)=O

InChI

1S/ClHO4.Li/c2-1(3,4)5;/h(H,2,3,4,5);/q;+1/p-1

InChI key

MHCFAGZWMAWTNR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Lithium perchlorate is a white salt that easily dissolves in water. It is commonly employed as an electrolytic salt in batteries due to its excellent electrical impedance, conductivity, hygroscopicity, and anodic stability properties. These characteristics are crucial for its role in battery applications. In addition, it is as a strong oxidizing agent and hence is used as an oxidizer in solid rocket propellants and organic reactions.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more information.

Application

Lithium perchlorate can be used:
  • As a precursor to prepare solid polymer electrolytes for rechargeable Li-ion batteries.
  • As an oxidizer to prepare polymer-based solid propellants.
  • To fabricate cobalt sulfide (CoS)-based counter electrode for dye-sensitized solar cells(DSSC).
Our Lithium perchlorate, with a 99.99% trace metals basis, possesses specific specifications that make it ideal for Lithium-ion batteries. It has low levels of anion traces such as chloride (Cl-): ≤0.003%and sulfate (SO42-): ≤0.001%. Additionally, it contains less than 5 ppm of heavy metals, further enhancing its suitability as an electrolytic salt for Lithium-ion batteries.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Ox. Sol. 2 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lithium garnet based free-standing solid polymer composite membrane for rechargeable lithium battery
K Karthik and Ramaswamy Murugan
Journal of Solid State Electrochemistry, 22, 2989-2998 (2018)
Lithium perchlorate-assisted electrodeposition of CoS catalyst surpassing the performance of platinum in dye sensitized solar cell
Mahir Gulen
Journal of Materials Science, 57, 3513-3524 (2022)
Kawamura, M. Kobayashi, S.
Tetrahedron Letters, 40, 1539-1539 (1999)
S Selvasekarapandian et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 65(5), 1234-1240 (2006-07-11)
The polymer electrolytes composed of poly(vinyl acetate) (PVAc) with various stoichiometric ratios of lithium perchlorate (LiClO(4)) salt have been prepared by solution casting method. The techniques Fourier transform infra-red (FTIR) and Laser Raman spectroscopy have been used to monitor polymer-salt
Increased T(c) in electrolyte-gated cuprates.
Anoop Singh Dhoot et al.
Advanced materials (Deerfield Beach, Fla.), 22(23), 2529-2533 (2010-05-07)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service