Skip to Content
Merck
All Photos(2)

Key Documents

809969

Sigma-Aldrich

Propylene carbonate

battery grade, ≥99%, acid <10 ppm, H2O <10 ppm

Synonym(s):

PC, 1,2-Propanediol cyclic carbonate, 4-Methyl-1,3-dioxolan-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6O3
CAS Number:
Molecular Weight:
102.09
Beilstein:
107913
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.23

grade

battery grade

Quality Level

vapor pressure

0.13 mmHg ( 20 °C)
0.98 mmHg ( 50 °C)

Assay

≥99%

form

liquid

autoignition temp.

851 °F

expl. lim.

14.3 %

impurities

≤10 ppm H2O
≤10 ppm acid

refractive index

n20/D 1.421 (lit.)

pH

7 (20 °C, 200 g/L)

bp

240 °C (lit.)

mp

−55 °C (lit.)

density

1.204 g/mL at 25 °C (lit.)
1.2 g/cm3

application(s)

battery manufacturing

SMILES string

CC1COC(=O)O1

InChI

1S/C4H6O3/c1-3-2-6-4(5)7-3/h3H,2H2,1H3

InChI key

RUOJZAUFBMNUDX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Alkyl carbonates find applications as solvents for lithium ion battery electrolytes and the use of high quality battery grade electrolytes having extremely low water (<10 ppm) and acid (<10 ppm) contents are critical for achieving high electrochemical performance.

Preparation Note

These additives have low water content (less than 100 ppm). Please handle under inert and moisture free enviornment (glove box). Keep containers tightly closed. Keep away from heat and ignition sources. Store in a cool and dry place. Avoid storing together with oxidizers.

Legal Information

Product of MU Ionic Solutions Corp

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

275.0 °F - closed cup

Flash Point(C)

135 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gor YG, et al.
Journal of Power Sources, 294 (2015)
Shi C, et al.
Electrochimica Acta, 174 (2015)

Articles

Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.

Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.

Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.

Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service