Skip to Content
Merck
All Photos(1)

Key Documents

63068

Sigma-Aldrich

Magnesium chloride hexahydrate

BioUltra, for molecular biology, ≥99.0% (KT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgCl2 · 6H2O
CAS Number:
Molecular Weight:
203.30
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.26
Assay:
≥99.0% (KT)
grade:
for molecular biology
form:
solid
solubility:
H2O: 1 M at 20 °C, clear, colorless
Pricing and availability is not currently available.

grade

for molecular biology

Quality Level

product line

BioUltra

Assay

≥99.0% (KT)

form

solid

impurities

DNases, none detected
RNases, none detected
insoluble matter, passes filter test
phosphatases, none detected
proteases, none detected
≤0.0002% total nitrogen (N)

pH

5.0-7.5 (25 °C, 1 M in H2O)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

phosphate (PO43-): ≤5 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤50 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤2000 mg/kg
Li: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤3000 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: 0.030
λ: 280 nm Amax: 0.025

SMILES string

O.O.O.O.O.O.Cl[Mg]Cl

InChI

1S/2ClH.Mg.6H2O/h2*1H;;6*1H2/q;;+2;;;;;;/p-2

InChI key

DHRRIBDTHFBPNG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Magnesium chloride hexahydrate has been used as a component of:
  • alkaline tris buffer to prepare labeling solution and to wash zebrafish embryos
  • HEPES-Tyrode solution for imaging cardiomyocyte cells
  • respiration solution, adenosine diphosphate (ADP) stock solution and wash solution to assess the respiration of permeabilized cardiomyocytes from mice

Caution

may agglomerate

Other Notes

Activation of restriction endonucleases[1]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J E Brooks
Methods in enzymology, 152, 113-129 (1987-01-01)
It is clear that we have still not exhausted all the restriction endonuclease specificities to be found in nature. Recently discovered BsmI is the first endonuclease recognizing a nonpalindromic sequence that cleaves within the site. Certainly other endonucleases belonging to
Günther Gratzl et al.
Colloids and surfaces. B, Biointerfaces, 126, 98-105 (2014-12-30)
The availability of polymeric antimicrobially active surfaces, which are mainly based on cationic surface effects, is limited. We have previously reported the discovery that, in addition to cationic surfaces, anionic surfaces based on poly(acrylic acid) (PAA) copolymers have a bactericidal
Monica Mazzarino et al.
Analytica chimica acta, 837, 70-82 (2014-07-09)
This article describes a liquid chromatographic/tandem mass spectrometric method, based on the use of precursor ion scan as the acquisition mode, specifically developed to detect indole-derived cannabinoids (phenylacetylindoles, naphthoylindoles and benzoylindoles) in biological fluids (saliva, urine and blood). The method
Carlos Sanchez-Martin et al.
Antioxidants & redox signaling, 34(7), 505-516 (2020-05-23)
Aims: TNF receptor-associated protein 1 (TRAP1), the mitochondrial paralog of the heat shock protein 90 (Hsp90) family of molecular chaperones, is required for neoplastic growth in several tumor cell models, where it inhibits succinate dehydrogenase (SDH) activity, thus favoring bioenergetic
Liang Yang et al.
International journal of pharmaceutics, 471(1-2), 258-263 (2014-05-20)
Telmisartan (TEL) belongs to BCS class II (low solubility/high permeability) and exhibits the pH-dependent drug release. Since 3-aminopropyl functionalized magnesium phyllosilicate (aminoclay) can intercalate or adsorb the negatively charged molecules via the electrostatic interaction, TEL-aminoclay complex was synthesized to improve

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service