Skip to Content
Merck
All Photos(1)

Documents

13152

Sigma-Aldrich

Magnesium chloride hexahydrate

puriss., meets analytical specification of Ph. Eur., BP, FCC, E511, 99-101%, ≤0.0001% Al

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgCl2 · 6H2O
CAS Number:
Molecular Weight:
203.30
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

puriss.

Quality Level

Assay

99-101%

form

solid

quality

meets analytical specification of Ph. Eur., BP, FCC, E511

impurities

acidity or alkalinity, complies
residual solvents, complies
≤0.0001% Al
≤0.005% ammonium (NH4)
≤0.01% free alkali (as MgO)
≤0.02% free acid (as HCl)
51.0-55.0% water (Karl Fischer)

anion traces

bromide (Br-): ≤500 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

Al: ≤1 mg/kg
As: ≤2 mg/kg
Ba: ≤20 mg/kg
Ca: ≤1000 mg/kg
Cu: ≤10 mg/kg
Fe: ≤5 mg/kg
K: ≤500 mg/kg
Na: ≤5000 mg/kg
Pb: ≤4 mg/kg
Zn: ≤10 mg/kg

suitability

complies for appearance of solution

SMILES string

O.O.O.O.O.O.Cl[Mg]Cl

InChI

1S/2ClH.Mg.6H2O/h2*1H;;6*1H2/q;;+2;;;;;;/p-2

InChI key

DHRRIBDTHFBPNG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Magnesium chloride hexahydrate is a colorless, crystalline solid. On heating, it loses water of crystallization and affords magnesium oxide. It has been tested as a potential phase change material (PCM) for use in cooking indoors and during the low intensity solar radiation periods. Due to its suitable thermo-physical properties, it can be used as storage media in solar cookers. Its flame retardancy action on 100% woven cotton fabric has been investigated. Its heat capacity has been evaluated.

Application

Magnesium chloride hexahydrate may be employed as a catalyst for the synthesis of 3,4-dihydropyrimidin-2-(1H)-one, via Biginelli condensation of aldehyde, 1,3-dicarbonyl compound and urea.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

One thousand thermal cycles of magnesium chloride hexahydrate as a promising PCM for indoor solar cooking.
El-Sebaii AA, et al.
Energy Conversion and Management , 52(4), 1771-1777 (2011)
Study of magnesium chloride hexahydrate as heat storage material.
Pilar R, et al.
Thermochimica Acta, 546, 81-86 (2012)
Magnesium Chloride Hexahydrate Catalyzed One-Pot Synthesis of 3, 4-Dihydropyrimidin-2-(1 H) ones Under Solvent-Free Conditions.
Zhang GL and Cai XH.
Synthetic Communications, 35(6), 829-833 (2005)
Thermal decomposition pathway of a cellulosic fabric impregnated by magnesium chloride hexahydrate as a flame-retardant.
Mostashari SM and Moafi HF.
Journal of Thermal Analysis and Calorimetry, 93(2), 589-594 (2008)
Liang Yang et al.
International journal of pharmaceutics, 471(1-2), 258-263 (2014-05-20)
Telmisartan (TEL) belongs to BCS class II (low solubility/high permeability) and exhibits the pH-dependent drug release. Since 3-aminopropyl functionalized magnesium phyllosilicate (aminoclay) can intercalate or adsorb the negatively charged molecules via the electrostatic interaction, TEL-aminoclay complex was synthesized to improve

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service