Skip to Content
Merck
All Photos(3)

Key Documents

S3014

Sigma-Aldrich

Sodium chloride

for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)

Synonym(s):

Halite, NaCl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaCl
CAS Number:
Molecular Weight:
58.44
Beilstein:
3534976
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.25
Assay:
≥99% (titration)
grade:
for molecular biology
form:
crystalline
solubility:
H2O: 1 g/10 mL, clear, colorless

grade

for molecular biology

Quality Level

Assay

≥99% (titration)

form

crystalline

impurities

DNAse, none detected
Endonuclease, none detected
NICKase, none detected
RNAse, none detected
protease, none detected

turbidity

≤3.5 NTU

pH

7

mp

801 °C (lit.)

solubility

H2O: 1 g/10 mL, clear, colorless

density

2.16 g/cm3 at 25 °C (77 °F)

anion traces

phosphate (PO43-): ≤5 ppm

cation traces

K: ≤50 ppm
≤5 ppm (Heavy Metals by ICP-OES)

suitability

suitable for Western blot
suitable for mass spectrometry
suitable for molecular biology

application(s)

cell analysis

foreign activity

DNase, RNase, and protease, none detected

SMILES string

[Na+].[Cl-]

InChI

1S/ClH.Na/h1H;/q;+1/p-1

InChI key

FAPWRFPIFSIZLT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

This product is designated as Molecular Biology grade and is suitable for molecular biology applications. It has been analyzed for the absence of nucleases and proteases.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jules D P Valentin et al.
Journal of colloid and interface science, 552, 247-257 (2019-05-28)
Many materials used in the medical settings such as catheters and contact lenses as well as most biological tissues are not purely elastic, but rather viscoelastic. While substrate elasticity has been investigated for its influence on bacterial adhesion, the impact
Zlatko Kopecki et al.
Oncotarget, 6(34), 36426-36440 (2015-10-27)
Flightless I (Flii) is an actin remodeling protein that affects cellular processes including adhesion, proliferation and migration. In order to determine the role of Flii during carcinogenesis, squamous cell carcinomas (SCCs) were induced in Flii heterozygous (Flii+/-), wild-type and Flii
Ram S Puranam et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 35(23), 8866-8881 (2015-06-13)
We identified a family in which a translocation between chromosomes X and 14 was associated with cognitive impairment and a complex genetic disorder termed "Genetic Epilepsy and Febrile Seizures Plus" (GEFS(+)). We demonstrate that the breakpoint on the X chromosome
Avanti Gokhale et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 35(19), 7643-7653 (2015-05-15)
Dysbindin is a schizophrenia susceptibility factor and subunit of the biogenesis of lysosome-related organelles complex 1 (BLOC-1) required for lysosome-related organelle biogenesis, and in neurons, synaptic vesicle assembly, neurotransmission, and plasticity. Protein networks, or interactomes, downstream of dysbindin/BLOC-1 remain partially
Meriam Ayadi et al.
Oncotarget, 6(21), 18518-18533 (2015-06-05)
Most solid tumors contain a subfraction of cells with stem/progenitor cell features. Stem cells are naturally chemoresistant suggesting that chronic chemotherapeutic stress may select for cells with increased "stemness". We carried out a comprehensive molecular and functional analysis of six

Protocols

Annealing is the process of heating and cooling two single-stranded oligonucleotides with complementary sequences.

Annealing is the process of heating and cooling two single-stranded oligonucleotides with complementary sequences.

Annealing is the process of heating and cooling two single-stranded oligonucleotides with complementary sequences.

Annealing is the process of heating and cooling two single-stranded oligonucleotides with complementary sequences.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service