63064
Magnesium chloride hexahydrate
BioUltra, ≥99.0% (KT)
Synonym(s):
MgCl2.6H2O, dichloromangesium hexahydrate
About This Item
Recommended Products
grade
reagent grade
Quality Level
product line
BioUltra
Assay
≥99.0% (KT)
form
solid
impurities
insoluble matter, passes filter test
≤0.0002% total nitrogen (N)
pH
5.0-7.5 (25 °C, 1 M in H2O)
solubility
H2O: 1 M at 20 °C, clear, colorless
anion traces
phosphate (PO43-): ≤5 mg/kg
sulfate (SO42-): ≤50 mg/kg
cation traces
Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤50 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤2000 mg/kg
Li: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤3000 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg
λ
1 M in H2O
UV absorption
λ: 260 nm Amax: 0.030
λ: 280 nm Amax: 0.025
SMILES string
O.O.O.O.O.O.Cl[Mg]Cl
InChI
1S/2ClH.Mg.6H2O/h2*1H;;6*1H2/q;;+2;;;;;;/p-2
InChI key
DHRRIBDTHFBPNG-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Application
- Magnesium chloride hexahydrate has been used in wash buffer for the purification of protein from Saccharomyces cerevisiae.
- It has been used for the preparation of extracellular (Ringer) solution for juxtacellular recording of single neurons in freely moving rats.
- It has been used as a supplement to study effect of desiccation on bacterial viability.
Other Notes
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service