Skip to Content
Merck
All Photos(1)

Key Documents

15614

Sigma-Aldrich

Hexamethylenetetramine

meets analytical specification of Ph. Eur., BP, 99-100.5% (calc. to the dried substance)

Synonym(s):

1,3,5,7-Tetraazatricyclo[3.3.1.13,7]decane, Hexamine, Methenamine, Urotropine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12N4
CAS Number:
Molecular Weight:
140.19
Beilstein:
2018
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.21

vapor pressure

<0.01 mmHg ( 20 °C)

Quality Level

Assay

99-100.5% (calc. to the dried substance)

form

solid

autoignition temp.

770 °F

quality

meets analytical specification of Ph. Eur., BP

impurities

acidic and alkalic reac. substances, complies
residual solvents, in accordance
≤0.002% heavy metals (as Pb)
≤0.005% free formaldehyde

loss

≤2% loss on drying

mp

280 °C (subl.) (lit.)

solubility

H2O: soluble

anion traces

chloride (Cl-): ≤100 mg/kg
sulfate (SO42-): ≤100 mg/kg

suitability

complies for appearance of solution

application(s)

pharmaceutical (small molecule)

SMILES string

C1N2CN3CN1CN(C2)C3

InChI

1S/C6H12N4/c1-7-2-9-4-8(1)5-10(3-7)6-9/h1-6H2

InChI key

VKYKSIONXSXAKP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Sol. 2 - Skin Sens. 1B

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 1

Flash Point(F)

482.0 °F - closed cup

Flash Point(C)

250 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E M Rodríguez et al.
Histochemistry, 81(3), 253-263 (1984-01-01)
The use of butyl-methyl-methacrylate embedding and the application of the silver methenamine (SM) method as a poststaining of the immunoperoxidase-DAB (IP) procedure led to the standardization of several useful methods for the visualization of tissue antigens at the light and
Enhanced quantum dot deposition on ZnO nanorods for photovoltaics through layer-by-layer processing.
<BIG>Joe Briscoe</BIG>
Journal of Materials Chemistry, 2517-2523 (2011)
Jianguo Hu et al.
Talanta, 93, 345-349 (2012-04-10)
A green, one-step method for synthesis of graphene-Au nanoparticles (graphene-AuNPs) was introduced in this article, using an environmentally benign hexamethylenetetramine (HMTA) as reducing and stabilizing agent. HMTA slowly was hydrolyzed to generate aldehyde ammonia to reduce graphene oxides (GO) and
Ann Pontén et al.
Contact dermatitis, 68(1), 50-53 (2012-10-06)
The currently used patch test concentration for formaldehyde is 1.0% (wt/vol) in water. However, clinical experience and previous studies suggest that 1.0% might be insufficient for detecting an optimized number of clinically relevant cases of contact allergy to formaldehyde. To
Vincent Consonni et al.
ACS nano, 8(5), 4761-4770 (2014-04-12)
Controlling the polarity of ZnO nanowires in addition to the uniformity of their structural morphology in terms of position, vertical alignment, length, diameter, and period is still a technological and fundamental challenge for real-world device integration. In order to tackle

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service