Skip to Content
Merck
All Photos(2)

Key Documents

52097

Supelco

Xylenol Orange disodium salt

for spectrophotometric determination of metal ions

Synonym(s):

3,3′-Bis[N,N-bis(carboxymethyl)aminomethyl]-o-cresolsulfonephthalein disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H30N2Na2O13S
CAS Number:
Molecular Weight:
716.62
UNSPSC Code:
12000000
NACRES:
NA.21

form

solid

Quality Level

quality

for spectrophotometric determination of metal ions

technique(s)

UV/Vis spectroscopy: suitable

SMILES string

[Na].[Na].Cc1cc(cc(CN(CC(O)=O)CC(O)=O)c1O)C2(OS(=O)(=O)c3ccccc23)c4cc(C)c(O)c(CN(CC(O)=O)CC(O)=O)c4

InChI

1S/C31H32N2O13S/c1-17-7-21(9-19(29(17)42)11-32(13-25(34)35)14-26(36)37)31(23-5-3-4-6-24(23)47(44,45)46-31)22-8-18(2)30(43)20(10-22)12-33(15-27(38)39)16-28(40)41/h3-10,42-43H,11-16H2,1-2H3,(H,34,35)(H,36,37)(H,38,39)(H,40,41)

InChI key

ORZHVTYKPFFVMG-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manoj K Semwal et al.
Journal of applied clinical medical physics, 9(4), 2809-2809 (2008-11-21)
We investigated the ferrous sulfate-benzoic acid-xylenol orange (FBX) aqueous chemical dosimeter for measurement of virtual (dynamic) wedge profiles on a linear accelerator. The layout for irradiation of the FBX-filled tubes mimicked a conventional linear detector array geometry. A comparison of
Lanlei Wang et al.
Clinical oral investigations, 15(6), 983-991 (2010-08-11)
The purpose of the present study was to evaluate histologically in beagle dogs the healing in acute dehiscence type defects following treatment with open flap debridement (OFD) with or without porous biphasic calcium phosphate (PBCP). Alveolar bone dehiscence defects were
Kenji Fukuzawa et al.
Journal of nutritional science and vitaminology, 55(1), 9-14 (2009-04-09)
The ferric-xylenol orange (FOX) method for measurement of hydroperoxides is based on a technique that employs reduction of peroxides in an acidic condition by Fe2+ and formation of the colored ferric-xylenol orange (XO/Fe3+) product with a peak at 560 nm.
Paul R T Kuzyk et al.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society, 28(4), 474-480 (2009-10-28)
The aim of this study was to compare angiogenesis and osteogenesis occurring within 8.0 mm diaphyseal defects created in canine tibiae treated using autograft or a biodegradable bone scaffold. All tibiae were reamed to 7.0 mm and fixed with a
Ya-Qin Zhao et al.
Journal of fluorescence, 22(1), 485-494 (2011-09-29)
Centrin, an EF-hand calcium-binding protein with high homology to calmodulin (CaM), is an essential component of microtubule-organizing center (MTOC). Lanthanide (Ln) ions can improve the stability, increase the amount and enhance the orderliness of microtubules, which are components of cytoskeleton.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service