Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Hexamethylenetetramine
Synonym(s):
Hexamethylenetetramine, 1,3,5,7-Tetraazatricyclo[3.3.1.1
3,7
]decane, Hexamine, Methenamine, Urotropine
Empirical Formula (Hill Notation):
C
6
H
12
N
4
CAS Number:
100-97-0
Molecular Weight:
140.19
EC Number:
202-905-8
Products
Product Category
building blocks (4)
Brand
Sigma-Aldrich (6)
Color
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Mode of Action
Special Grade
ACS reagent (2)
Anhydrous (1)
Puriss P.A. (1)
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
8.18712
Hexamethylenetetramine,
for synthesis
Expand
View Pricing
398160
Hexamethylenetetramine,
ACS reagent, ≥99.0%
Expand
View Pricing
H11300
Hexamethylenetetramine,
ReagentPlus
®
, 99%
Expand
View Pricing
33233
Hexamethylenetetramine,
puriss. p.a., reag. Ph. Eur., ≥99.5% (calc. to the dried substance)
Expand
View Pricing
13-0750
Hexamethylenetetramine,
JIS special grade, ≥99.0%
Expand
View Pricing
797979
Hexamethylenetetramine,
anhydrous, free-flowing, Redi-Dri
™
, ACS reagent, ≥99.0%
Expand
View Pricing
13-0760
Hexamethylenetetramine,
SAJ first grade, ≥98.5%
Expand
View Pricing