Skip to Content
Merck
All Photos(4)

Key Documents

398187

Sigma-Aldrich

Xylenol Orange tetrasodium salt

ACS reagent

Synonym(s):

3,3′-Bis[N,N-bis(carboxymethyl)aminomethyl]-o-cresolsulfonephthalein tetrasodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H28N2Na4O13S
CAS Number:
Molecular Weight:
760.58
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

form

powder or crystals

technique(s)

titration: suitable

color

very dark brown to very dark red

mp

195 °C (dec.) (lit.)

λmax

580 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].[Na+].[Na+].Cc1cc(cc(CN(CC([O-])=O)CC([O-])=O)c1O)C2(OS(=O)(=O)c3ccccc23)c4cc(C)c(O)c(CN(CC([O-])=O)CC([O-])=O)c4

InChI

1S/C31H32N2O13S.4Na/c1-17-7-21(9-19(29(17)42)11-32(13-25(34)35)14-26(36)37)31(23-5-3-4-6-24(23)47(44,45)46-31)22-8-18(2)30(43)20(10-22)12-33(15-27(38)39)16-28(40)41;;;;/h3-10,42-43H,11-16H2,1-2H3,(H,34,35)(H,36,37)(H,38,39)(H,40,41);;;;/q;4*+1/p-4

InChI key

YIIQUYHVWRJXEM-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

General description

Xylenol Orange tetrasodium salt is a fluorochrome acid dye with complexing and chelating properties that make it useful as an indicator in the spectrophotometric detection and assay of metal ions such as calcium, cadmium, bismuth, zinc, lead, and mercury. Its tetrasodium salt especially is used as an indicator in metal titrations. It changes color from yellow to red between pH 6.4 to 10.4. It has six potentially anionic substituents, one being sulfonate, and is anionic under staining conditions and during most laboratory applications.[1][2]

Application

  • Xylenol Orange has been used as a vital stain to study calcification in bone development, injury, and repair.[1]
  • It has been used in the fluorescent histochemical detection of alkaline phosphatase.[1]
  • It has been used in FBX (ferrous benzoic xylenol orange) gel preparation to study the effects of MRI (magnetic resonance imaging) parameters on image quality.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Use of xylenol orange and cetylpyridinium chloride in rapid spectrophotometric determination of zinc in pharmaceutical products
M Benamor , K Belhamel, M T Draa
Journal of Pharmaceutical and Biomedical Analysis, 23 (6) (2000)
Rost F W D
Fluorescence Microscopy, 2 (1992)
Conn?s Biological Stains: A Handbook of Dyes, Stains and Fluorochromes for Use in Biology and Medicine (10th ed.)
R. W. Horobin and J. A. Kiernan (Eds.).
Conn?s Biological Stains (2002)
Aalia Nazir et al.
JPMA. The Journal of the Pakistan Medical Association, 60(6), 470-473 (2010-06-10)
This study focuses on the diverse effects of MRI parameters on image quality using widely available imaging pulse sequences. A tissue equivalent gel system has been used for Magnetic Resonance Imaging by using Xylenol orange dye with Fricke-Benzoic solution which

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service