Skip to Content
Merck
All Photos(2)

Key Documents

01514

Supelco

[8]-Gingerol

analytical standard

Synonym(s):

(S)-5-Hydroxy-1-(4-hydroxy-3-methoxyphenyl)-3-dodecanone, 8-Gingerol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H30O4
CAS Number:
Molecular Weight:
322.44
Beilstein:
2059802
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

CCCCCCC[C@H](O)CC(=O)CCc1ccc(O)c(OC)c1

InChI

1S/C19H30O4/c1-3-4-5-6-7-8-16(20)14-17(21)11-9-15-10-12-18(22)19(13-15)23-2/h10,12-13,16,20,22H,3-9,11,14H2,1-2H3/t16-/m0/s1

InChI key

BCIWKKMTBRYQJU-INIZCTEOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Gingerol is the pungent component of ginger roots. 8-Gingerol has a chain length of n8. It is a homologous series of phenolic ketones.
[8]-Gingerol is a bioactive constituent of ginger. It exhibits anti-inflammatory, anticancer, anti-oxidant and anti-ulcer properties.

Application

It may be used as reference standard during quantification of 6-, 8- and 10-gingerols and 6-shogaol in human plasma using HPLC with electrochemical detection.
[8]-Gingerol may be used as a reference standard in the determination of [8]-gingerol in dietary supplements, food products and beverages using high-performance liquid chromatography (HPLC).

Biochem/physiol Actions

[8]-Gingerol is a bioactive compound found in ginger (Zingiber officinale) with anti-inflammatory and antioxidant activity.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: zingiber

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-performance liquid chromatographic analysis of 6-gingerol, 8-gingerol, 10-gingerol, and 6-shogaol in ginger-containing dietary supplements, spices, teas, and beverages.
Schwertner HA and Rios DC
Journal of Chromatography. B, Biomedical Sciences and Applications, 856(1-2), 41-47 (2007)
Suzanna M Zick et al.
International journal of biomedical science : IJBS, 6(3), 233-240 (2010-11-13)
Zingiber officinale is one of the most commonly used spices. We developed a method to determine the main pungent ginger constituents, 6-, 8- and 10-gingerols and 6-shogaol in human plasma. Quantitation was achieved using a reversed-phase C(18) column using high-performance
Comparative antioxidant and anti-inflammatory effects of [6]-gingerol,[8]-gingerol,[10]-gingerol and [6]-shogaol.
Dugasani S, et al.
Journal of Ethnopharmacology, 127(2), 515-520 (2010)
High-performance liquid chromatography-electrospray mass spectrometric analysis of pungent constituents of ginger.
He, Xian-guo, et al.
Journal of Chromatography A, 796.2, 327-334 (1998)
Andreas Nievergelt et al.
Bioorganic & medicinal chemistry, 18(9), 3345-3351 (2010-04-07)
Animal studies suggest that ginger (Zingiber officinale Roscoe) reduces anxiety. In this study, bioactivity-guided fractionation of a ginger extract identified nine compounds that interact with the human serotonin 5-HT(1A) receptor with significant to moderate binding affinities (K(i)=3-20 microM). [(35)S]-GTP gamma

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service