Skip to Content
Merck
All Photos(1)

Key Documents

G5923

Sigma-Aldrich

[8]-Gingerol

Synonym(s):

(S)-5-Hydroxy-1-(4-hydroxy-3-methoxyphenyl)-3-dodecanone, 8-Gingerol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H30O4
CAS Number:
Molecular Weight:
322.44
Beilstein:
2059802
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

color

white to yellow

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

CCCCCCC[C@H](O)CC(=O)CCc1ccc(O)c(OC)c1

InChI

1S/C19H30O4/c1-3-4-5-6-7-8-16(20)14-17(21)11-9-15-10-12-18(22)19(13-15)23-2/h10,12-13,16,20,22H,3-9,11,14H2,1-2H3/t16-/m0/s1

InChI key

BCIWKKMTBRYQJU-INIZCTEOSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

[8]-Gingerol is a bioactive compound found in ginger (Zingiber officinale) with anti-inflammatory and antioxidant activity.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andreas Nievergelt et al.
Bioorganic & medicinal chemistry, 18(9), 3345-3351 (2010-04-07)
Animal studies suggest that ginger (Zingiber officinale Roscoe) reduces anxiety. In this study, bioactivity-guided fractionation of a ginger extract identified nine compounds that interact with the human serotonin 5-HT(1A) receptor with significant to moderate binding affinities (K(i)=3-20 microM). [(35)S]-GTP gamma
Zhufeng Wu et al.
The Journal of pharmacy and pharmacology, 67(4), 583-596 (2014-12-17)
To determine the reaction kinetics for regioselective glucuronidation of gingerols (i.e. 6-, 8- and 10-gingerol) by human liver microsomes and expressed UDP-glucuronosyltransferase (UGT) enzymes, and to identify the main UGT enzymes involved in regioselective glucuronidation of gingerols. The rates of
Wen Xu et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 986-987, 69-84 (2015-02-25)
Gua-Lou-Gui-Zhi decoction (GLGZD) is a classical formula of traditional Chinese medicine, which has been commonly used to treat dysfunction after stroke, epilepsy and spinal cord injury. In this study, a systematic method was established for chemical profiling and quantification analysis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service