Skip to Content
Merck
All Photos(1)

Key Documents

O1514

Sigma-Aldrich

Olive oil

highly refined, low acidity

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

form

liquid

Quality Level

quality

highly refined, low acidity

density

D20/4, 0.895-0.924

functional group

ester
oleic acid

lipid type

oils

shipped in

ambient

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Olive oil is rich in the phenolic component, squalene glucoside. The extra-virgin olive oil (VOQ) is antioxidant rich and a potential reactive oxygen scavenger.[1] Olive oil is rich in monounsaturated long-chain fatty acids and is not prone to oxidation. Olive oil impairs xanthine oxidase activity and could have potential in modulating tumor progression.[2] The biophenols in olive oil may elicit protection in metabolic disorders, cardiovascular diseases and neurodegeneration.[3]

Application

Olive oil has been used as an oral supplement for metabolic phenotyping in mice [4]. It has been used for dissolution of vitamin E[5], whey protein[6]and drugs.[7]

Biochem/physiol Actions

The health benefits of olive oil result from its higher proportion of monounsaturated fats, as well as its antioxidant constituents: hydroxytyrosol, oleuropein and oleocanthal.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

The antioxidant/anticancer potential of phenolic compounds isolated from olive oil
Owen RW, et al.
European Journal of Cancer, 36(10), 1235-1247 (2000)
Dietary protein dilution limits dyslipidemia in obesity through FGF21-driven fatty acid clearance
Maida A, et al.
The Journal of Nutritional Biochemistry, 57(6), 189-196 (2018)
Ginseng extract reduces tacrolimus-induced oxidative stress by modulating autophagy in pancreatic beta cells
Lim SW, et al.
Laboratory Investigation; a Journal of Technical Methods and Pathology, 97(11), 1271-1271 (2017)
Pharma-nutritional properties of olive oil phenols. transfer of new findings to human nutrition
Crespo M, et al.
Foods (Basel, Switzerland), 7(6), 90-90 (2018)
Kyung-Taek Rim et al.
Safety and health at work, 6(3), 184-191 (2016-03-02)
Chemical mutagenicity is a major hazard that is important to workers' health. Despite the use of large amounts of allyl chloride, the available mutagenicity data for this chemical remains controversial. To clarify the mutagenicity of allyl chloride and because a

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service