Skip to Content
Merck
All Photos(2)

Key Documents

42630

Supelco

[10]-Gingerol

analytical standard

Synonym(s):

(S)- 5-Hydroxy-1-(4-hydroxy-3-methoxyphenyl)-3-tetradecanone, 10-Gingerol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H34O4
CAS Number:
Molecular Weight:
350.49
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥94.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CCCCCCCCC[C@H](O)CC(=O)CCc1ccc(O)c(OC)c1

InChI

1S/C21H34O4/c1-3-4-5-6-7-8-9-10-18(22)16-19(23)13-11-17-12-14-20(24)21(15-17)25-2/h12,14-15,18,22,24H,3-11,13,16H2,1-2H3/t18-/m0/s1

InChI key

AIULWNKTYPZYAN-SFHVURJKSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

[10]-Gingerol is a bioactive compound found in ginger (Zingiber officinale) with anti-inflammatory and antioxidant activity.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: zingiber

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

331.3 °F - closed cup

Flash Point(C)

166.3 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bing-Hung Chen et al.
Journal of natural products, 72(5), 950-953 (2009-03-11)
In the present study on five pure phenolic compounds (1-5) isolated from the rhizomes of Zingiber officinale (ginger) and investigated for their antiallergic potency, rat basophilic leukemia (RBL-2H3) cells were incubated with these compounds and the release of beta-hexosaminidase was
Andreas Nievergelt et al.
Bioorganic & medicinal chemistry, 18(9), 3345-3351 (2010-04-07)
Animal studies suggest that ginger (Zingiber officinale Roscoe) reduces anxiety. In this study, bioactivity-guided fractionation of a ginger extract identified nine compounds that interact with the human serotonin 5-HT(1A) receptor with significant to moderate binding affinities (K(i)=3-20 microM). [(35)S]-GTP gamma
Zhufeng Wu et al.
The Journal of pharmacy and pharmacology, 67(4), 583-596 (2014-12-17)
To determine the reaction kinetics for regioselective glucuronidation of gingerols (i.e. 6-, 8- and 10-gingerol) by human liver microsomes and expressed UDP-glucuronosyltransferase (UGT) enzymes, and to identify the main UGT enzymes involved in regioselective glucuronidation of gingerols. The rates of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service